Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48523909-48524140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBP
Entrez Gene ID
10682
Gene Name
emopamil binding protein (sterol isomerase)
Gene Aliases
CDPX2, CHO2, CPX, CPXD, MEND
UniGene
Hs.30619
Gene Chromosome Location
Chr.X: 48521776-48528716 on Build GRCh38
Transcripts
NM_006579.2

SNP Detail

SNP ID Chromosome Location
rs960238368 Chr.X: 48523945-48523945 on Build GRCh38
rs886038570 Chr.X: 48524077-48524077 on Build GRCh38
rs104894799 Chr.X: 48523958-48523958 on Build GRCh38
rs782103792 Chr.X: 48524099-48524099 on Build GRCh38
rs377038311 Chr.X: 48524060-48524060 on Build GRCh38
rs587783604 Chr.X: 48524063-48524067 on Build GRCh38
rs782558322 Chr.X: 48523946-48523946 on Build GRCh38
rs782294297 Chr.X: 48523965-48523965 on Build GRCh38
rs781836618 Chr.X: 48524091-48524091 on Build GRCh38
rs781855861 Chr.X: 48523943-48523943 on Build GRCh38
rs782366560 Chr.X: 48524020-48524020 on Build GRCh38
rs182712356 Chr.X: 48524053-48524053 on Build GRCh38
rs587783605 Chr.X: 48524070-48524070 on Build GRCh38
rs35265835 Chr.X: 48523964-48523965 on Build GRCh38
rs104894800 Chr.X: 48524009-48524009 on Build GRCh38
rs797045542 Chr.X: 48523974-48523975 on Build GRCh38
rs587783603 Chr.X: 48523989-48523989 on Build GRCh38
rs145509273 Chr.X: 48524032-48524032 on Build GRCh38
rs781993981 Chr.X: 48524028-48524028 on Build GRCh38
rs782615162 Chr.X: 48523956-48523956 on Build GRCh38
rs797045543 Chr.X: 48523996-48523997 on Build GRCh38
rs782780770 Chr.X: 48524120-48524120 on Build GRCh38
rs782217564 Chr.X: 48523957-48523957 on Build GRCh38
rs372974717 Chr.X: 48523955-48523955 on Build GRCh38
rs782638360 Chr.X: 48523959-48523959 on Build GRCh38
rs587783602 Chr.X: 48523985-48523985 on Build GRCh38
rs782185557 Chr.X: 48524008-48524008 on Build GRCh38
rs781800812 Chr.X: 48523947-48523947 on Build GRCh38
rs371430396 Chr.X: 48524104-48524104 on Build GRCh38
rs587783600 Chr.X: 48523953-48523953 on Build GRCh38
rs587783601 Chr.X: 48523975-48523975 on Build GRCh38
rs782807455 Chr.X: 48524033-48524033 on Build GRCh38
rs587783606 Chr.X: 48524074-48524074 on Build GRCh38
rs782423797 Chr.X: 48523979-48523979 on Build GRCh38
rs797045153 Chr.X: 48523995-48523995 on Build GRCh38
rs781939301 Chr.X: 48523990-48523990 on Build GRCh38

View Full Product Details