Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38619162-38619407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3K
Entrez Gene ID
27335
Gene Name
eukaryotic translation initiation factor 3 subunit K
Gene Aliases
ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001
UniGene
Hs.314359
Gene Chromosome Location
Chr.19: 38619072-38636959 on Build GRCh38
Transcripts
NM_001300992.1 NM_001308393.1 NM_013234.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524976 c.48G>A p.K16K Chr.19: 38619316-38619316 on Build GRCh38
7156331 c.13G>A p.E5K Chr.19: 38619281-38619281 on Build GRCh38
5649324 c.13G>C p.E5Q Chr.19: 38619281-38619281 on Build GRCh38
74496 c.52A>G p.I18V Chr.19: 38619320-38619320 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772662121 Chr.19: 38619335-38619335 on Build GRCh38
rs2232995 Chr.19: 38619222-38619222 on Build GRCh38
rs2232994 Chr.19: 38619221-38619221 on Build GRCh38
rs371960290 Chr.19: 38619263-38619263 on Build GRCh38
rs954636630 Chr.19: 38619250-38619250 on Build GRCh38
rs370051027 Chr.19: 38619338-38619338 on Build GRCh38
rs771747294 Chr.19: 38619273-38619274 on Build GRCh38
rs760094887 Chr.19: 38619345-38619345 on Build GRCh38
rs760228569 Chr.19: 38619338-38619341 on Build GRCh38
rs770871541 Chr.19: 38619347-38619359 on Build GRCh38
rs112447585 Chr.19: 38619201-38619201 on Build GRCh38
rs200004003 Chr.19: 38619363-38619363 on Build GRCh38
rs773991767 Chr.19: 38619308-38619308 on Build GRCh38
rs944712046 Chr.19: 38619301-38619301 on Build GRCh38
rs895379233 Chr.19: 38619196-38619196 on Build GRCh38
rs745802177 Chr.19: 38619274-38619274 on Build GRCh38
rs765788701 Chr.19: 38619349-38619349 on Build GRCh38
rs369039219 Chr.19: 38619248-38619248 on Build GRCh38
rs753286144 Chr.19: 38619351-38619351 on Build GRCh38
rs375807765 Chr.19: 38619241-38619241 on Build GRCh38
rs767541899 Chr.19: 38619195-38619195 on Build GRCh38
rs201812654 Chr.19: 38619219-38619219 on Build GRCh38
rs372381183 Chr.19: 38619224-38619224 on Build GRCh38
rs751734799 Chr.19: 38619244-38619244 on Build GRCh38
rs749037064 Chr.19: 38619300-38619300 on Build GRCh38
rs376805949 Chr.19: 38619337-38619337 on Build GRCh38
rs757488533 Chr.19: 38619369-38619369 on Build GRCh38
rs2232997 Chr.19: 38619335-38619335 on Build GRCh38
rs974578023 Chr.19: 38619229-38619229 on Build GRCh38
rs763310239 Chr.19: 38619354-38619354 on Build GRCh38
rs376864603 Chr.19: 38619330-38619330 on Build GRCh38
rs751793442 Chr.19: 38619366-38619366 on Build GRCh38
rs959668776 Chr.19: 38619192-38619192 on Build GRCh38
rs2232996 Chr.19: 38619304-38619304 on Build GRCh38
rs374142497 Chr.19: 38619336-38619336 on Build GRCh38
rs781301896 Chr.19: 38619373-38619373 on Build GRCh38
rs769764967 Chr.19: 38619279-38619279 on Build GRCh38
rs779805764 Chr.19: 38619299-38619299 on Build GRCh38
rs530039358 Chr.19: 38619225-38619225 on Build GRCh38
rs1050067345 Chr.19: 38619368-38619368 on Build GRCh38
rs1024970477 Chr.19: 38619207-38619207 on Build GRCh38

View Full Product Details