Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144475512-144475781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXH1
Entrez Gene ID
8928
Gene Name
forkhead box H1
Gene Aliases
FAST-1, FAST1
UniGene
Hs.708365
Gene Chromosome Location
Chr.8: 144473732-144476335 on Build GRCh38
Transcripts
NM_003923.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207421 c.55C>T p.Q19* Chr.8: 144475702-144475702 on Build GRCh38
6666246 c.29G>T p.G10V Chr.8: 144475728-144475728 on Build GRCh38
6367867 c.10T>A p.C4S Chr.8: 144475747-144475747 on Build GRCh38
5531401 c.30G>A p.G10G Chr.8: 144475727-144475727 on Build GRCh38
5531400 c.29G>A p.G10E Chr.8: 144475728-144475728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766946089 Chr.8: 144475679-144475679 on Build GRCh38
rs748058531 Chr.8: 144475703-144475703 on Build GRCh38
rs143968722 Chr.8: 144475615-144475615 on Build GRCh38
rs373519961 Chr.8: 144475695-144475695 on Build GRCh38
rs573656675 Chr.8: 144475759-144475759 on Build GRCh38
rs201224517 Chr.8: 144475655-144475655 on Build GRCh38
rs770133767 Chr.8: 144475602-144475602 on Build GRCh38
rs775978145 Chr.8: 144475734-144475734 on Build GRCh38
rs759694605 Chr.8: 144475594-144475594 on Build GRCh38
rs778783647 Chr.8: 144475699-144475699 on Build GRCh38
rs774860387 Chr.8: 144475744-144475744 on Build GRCh38
rs761332996 Chr.8: 144475667-144475667 on Build GRCh38
rs776865143 Chr.8: 144475592-144475592 on Build GRCh38
rs547103300 Chr.8: 144475544-144475544 on Build GRCh38
rs180724802 Chr.8: 144475710-144475710 on Build GRCh38
rs973588241 Chr.8: 144475626-144475626 on Build GRCh38
rs201691370 Chr.8: 144475627-144475627 on Build GRCh38
rs539395830 Chr.8: 144475652-144475652 on Build GRCh38
rs955149258 Chr.8: 144475577-144475577 on Build GRCh38
rs755788535 Chr.8: 144475682-144475682 on Build GRCh38
rs750023968 Chr.8: 144475680-144475680 on Build GRCh38
rs199791123 Chr.8: 144475607-144475607 on Build GRCh38
rs562065615 Chr.8: 144475555-144475555 on Build GRCh38
rs1043646541 Chr.8: 144475708-144475708 on Build GRCh38
rs201064389 Chr.8: 144475542-144475542 on Build GRCh38
rs768179507 Chr.8: 144475619-144475621 on Build GRCh38
rs139951652 Chr.8: 144475656-144475656 on Build GRCh38
rs1007629465 Chr.8: 144475717-144475717 on Build GRCh38
rs369306218 Chr.8: 144475635-144475635 on Build GRCh38
rs200335846 Chr.8: 144475725-144475725 on Build GRCh38
rs983462864 Chr.8: 144475639-144475639 on Build GRCh38
rs753680286 Chr.8: 144475685-144475685 on Build GRCh38
rs200913432 Chr.8: 144475588-144475588 on Build GRCh38
rs139312396 Chr.8: 144475659-144475659 on Build GRCh38
rs946590436 Chr.8: 144475700-144475700 on Build GRCh38
rs762230558 Chr.8: 144475752-144475752 on Build GRCh38
rs895247507 Chr.8: 144475688-144475688 on Build GRCh38
rs749719443 Chr.8: 144475554-144475555 on Build GRCh38
rs779809876 Chr.8: 144475683-144475683 on Build GRCh38
rs1029313710 Chr.8: 144475758-144475758 on Build GRCh38
rs781140141 Chr.8: 144475713-144475713 on Build GRCh38
rs909912152 Chr.8: 144475612-144475612 on Build GRCh38
rs769160861 Chr.8: 144475743-144475743 on Build GRCh38
rs374587860 Chr.8: 144475742-144475742 on Build GRCh38

View Full Product Details