Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41813313-41813544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC134
Entrez Gene ID
79879
Gene Name
coiled-coil domain containing 134
Gene Aliases
-
UniGene
Hs.474991
Gene Chromosome Location
Chr.22: 41800610-41832164 on Build GRCh38
Transcripts
NM_001304797.1 NM_024821.3 XM_005261748.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3061394 c.361C>T p.R121C Chr.22: 41813314-41813314 on Build GRCh38
4928131 c.470C>T p.P157L Chr.22: 41813423-41813423 on Build GRCh38
4943722 c.434G>C p.S145T Chr.22: 41813387-41813387 on Build GRCh38
5778509 c.468G>A p.G156G Chr.22: 41813421-41813421 on Build GRCh38
341242 c.398A>G p.N133S Chr.22: 41813351-41813351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753645777 Chr.22: 41813374-41813374 on Build GRCh38
rs781027539 Chr.22: 41813422-41813422 on Build GRCh38
rs754948072 Chr.22: 41813421-41813421 on Build GRCh38
rs530566441 Chr.22: 41813499-41813499 on Build GRCh38
rs376112163 Chr.22: 41813485-41813485 on Build GRCh38
rs373803096 Chr.22: 41813484-41813484 on Build GRCh38
rs755935696 Chr.22: 41813497-41813497 on Build GRCh38
rs751030130 Chr.22: 41813487-41813487 on Build GRCh38
rs200210669 Chr.22: 41813352-41813352 on Build GRCh38
rs373006626 Chr.22: 41813463-41813463 on Build GRCh38
rs757901016 Chr.22: 41813416-41813416 on Build GRCh38
rs866580795 Chr.22: 41813348-41813348 on Build GRCh38
rs753141912 Chr.22: 41813402-41813402 on Build GRCh38
rs746927683 Chr.22: 41813420-41813420 on Build GRCh38
rs760084688 Chr.22: 41813466-41813466 on Build GRCh38
rs551789392 Chr.22: 41813452-41813452 on Build GRCh38
rs373211102 Chr.22: 41813334-41813334 on Build GRCh38
rs748287551 Chr.22: 41813336-41813336 on Build GRCh38
rs376681838 Chr.22: 41813476-41813476 on Build GRCh38
rs531928294 Chr.22: 41813357-41813357 on Build GRCh38
rs374001821 Chr.22: 41813407-41813407 on Build GRCh38
rs370929512 Chr.22: 41813489-41813489 on Build GRCh38
rs766574164 Chr.22: 41813362-41813362 on Build GRCh38
rs771119365 Chr.22: 41813458-41813458 on Build GRCh38
rs780206116 Chr.22: 41813338-41813339 on Build GRCh38
rs376462390 Chr.22: 41813471-41813471 on Build GRCh38
rs760176243 Chr.22: 41813377-41813377 on Build GRCh38
rs773419482 Chr.22: 41813430-41813430 on Build GRCh38
rs768428856 Chr.22: 41813332-41813332 on Build GRCh38
rs982240473 Chr.22: 41813502-41813502 on Build GRCh38
rs767967796 Chr.22: 41813399-41813399 on Build GRCh38
rs148694139 Chr.22: 41813429-41813429 on Build GRCh38
rs775522013 Chr.22: 41813376-41813376 on Build GRCh38
rs762837401 Chr.22: 41813454-41813455 on Build GRCh38
rs747219908 Chr.22: 41813493-41813493 on Build GRCh38
rs377461039 Chr.22: 41813353-41813353 on Build GRCh38
rs768096527 Chr.22: 41813470-41813470 on Build GRCh38
rs764059828 Chr.22: 41813333-41813333 on Build GRCh38
rs765280336 Chr.22: 41813346-41813346 on Build GRCh38
rs145231436 Chr.22: 41813414-41813414 on Build GRCh38
rs747017405 Chr.22: 41813331-41813331 on Build GRCh38
rs542697204 Chr.22: 41813507-41813507 on Build GRCh38
rs371211408 Chr.22: 41813367-41813367 on Build GRCh38
rs765845212 Chr.22: 41813486-41813486 on Build GRCh38
rs781130462 Chr.22: 41813490-41813490 on Build GRCh38
rs756465884 Chr.22: 41813403-41813403 on Build GRCh38
rs749537268 Chr.22: 41813445-41813445 on Build GRCh38
rs368664191 Chr.22: 41813330-41813330 on Build GRCh38
rs779598906 Chr.22: 41813419-41813419 on Build GRCh38

View Full Product Details