Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 162151563-162151804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG2
Entrez Gene ID
2566
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases
CAE2, ECA2, GEFSP3
UniGene
Hs.7195
Gene Chromosome Location
Chr.5: 162067642-162155539 on Build GRCh38
Transcripts
NM_000816.3 NM_198903.2 NM_198904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2156243 c.1136G>A p.R379Q Chr.5: 162151737-162151737 on Build GRCh38
6668684 c.1152+9delT p.? Chr.5: 162151762-162151762 on Build GRCh38
4568638 c.1131T>A p.L377L Chr.5: 162151732-162151732 on Build GRCh38
207498 c.1135C>T p.R379W Chr.5: 162151736-162151736 on Build GRCh38
6470151 c.1137G>T p.R379R Chr.5: 162151738-162151738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988478980 Chr.5: 162151657-162151658 on Build GRCh38
rs575551694 Chr.5: 162151699-162151699 on Build GRCh38
rs374405346 Chr.5: 162151760-162151760 on Build GRCh38
rs757830756 Chr.5: 162151612-162151612 on Build GRCh38
rs959744569 Chr.5: 162151617-162151618 on Build GRCh38
rs758652747 Chr.5: 162151729-162151729 on Build GRCh38
rs750592610 Chr.5: 162151725-162151725 on Build GRCh38
rs943549919 Chr.5: 162151722-162151722 on Build GRCh38
rs755430911 Chr.5: 162151737-162151737 on Build GRCh38
rs557384687 Chr.5: 162151679-162151679 on Build GRCh38
rs780368645 Chr.5: 162151732-162151732 on Build GRCh38
rs1044397787 Chr.5: 162151669-162151669 on Build GRCh38
rs113115820 Chr.5: 162151668-162151668 on Build GRCh38
rs535591029 Chr.5: 162151659-162151659 on Build GRCh38
rs201128638 Chr.5: 162151705-162151705 on Build GRCh38
rs753854680 Chr.5: 162151708-162151708 on Build GRCh38
rs574546726 Chr.5: 162151593-162151593 on Build GRCh38
rs947064937 Chr.5: 162151658-162151658 on Build GRCh38
rs757290958 Chr.5: 162151709-162151709 on Build GRCh38
rs267600532 Chr.5: 162151746-162151746 on Build GRCh38
rs912927082 Chr.5: 162151658-162151658 on Build GRCh38
rs199574539 Chr.5: 162151742-162151742 on Build GRCh38
rs769292441 Chr.5: 162151773-162151773 on Build GRCh38
rs778869689 Chr.5: 162151717-162151717 on Build GRCh38
rs747426141 Chr.5: 162151736-162151736 on Build GRCh38
rs200677917 Chr.5: 162151712-162151712 on Build GRCh38
rs781756750 Chr.5: 162151744-162151744 on Build GRCh38

View Full Product Details