Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181538105-181538330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERKL
Entrez Gene ID
375298
Gene Name
ceramide kinase like
Gene Aliases
RP26
UniGene
Hs.732358
Gene Chromosome Location
Chr.2: 181536672-181657107 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1 NM_000885.5

Gene Symbol
ITGA4
Entrez Gene ID
3676
Gene Name
integrin subunit alpha 4
Gene Aliases
CD49D, IA4
UniGene
Hs.440955
Gene Chromosome Location
Chr.2: 181456892-181538928 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1 NM_000885.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3574772 c.1657G>A p.E553K Chr.2: 181538204-181538204 on Build GRCh38
2710423 c.1649G>A p.G550E Chr.2: 181538212-181538212 on Build GRCh38
4136749 c.1561C>A p.L521I Chr.2: 181538222-181538222 on Build GRCh38
2710426 c.1557C>G p.I519M Chr.2: 181538226-181538226 on Build GRCh38
2710425 c.1635C>G p.I545M Chr.2: 181538226-181538226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747579450 Chr.2: 181538191-181538192 on Build GRCh38
rs777753662 Chr.2: 181538159-181538160 on Build GRCh38
rs1030087021 Chr.2: 181538296-181538296 on Build GRCh38
rs750559730 Chr.2: 181538128-181538157 on Build GRCh38
rs752334385 Chr.2: 181538187-181538187 on Build GRCh38
rs1053928939 Chr.2: 181538252-181538252 on Build GRCh38
rs779379367 Chr.2: 181538190-181538191 on Build GRCh38
rs549186353 Chr.2: 181538145-181538145 on Build GRCh38
rs550012321 Chr.2: 181538130-181538131 on Build GRCh38
rs766512343 Chr.2: 181538269-181538281 on Build GRCh38
rs752704341 Chr.2: 181538149-181538150 on Build GRCh38
rs745449502 Chr.2: 181538183-181538184 on Build GRCh38
rs755313231 Chr.2: 181538239-181538239 on Build GRCh38
rs777027354 Chr.2: 181538218-181538219 on Build GRCh38
rs749423367 Chr.2: 181538264-181538264 on Build GRCh38
rs1005920378 Chr.2: 181538259-181538259 on Build GRCh38
rs910844553 Chr.2: 181538243-181538243 on Build GRCh38
rs753666698 Chr.2: 181538160-181538161 on Build GRCh38
rs747421880 Chr.2: 181538126-181538126 on Build GRCh38
rs972243538 Chr.2: 181538130-181538130 on Build GRCh38
rs755723305 Chr.2: 181538189-181538189 on Build GRCh38
rs762150607 Chr.2: 181538224-181538225 on Build GRCh38
rs755966802 Chr.2: 181538152-181538152 on Build GRCh38
rs769934349 Chr.2: 181538244-181538244 on Build GRCh38
rs565730415 Chr.2: 181538148-181538148 on Build GRCh38
rs751564367 Chr.2: 181538274-181538281 on Build GRCh38
rs778707393 Chr.2: 181538182-181538183 on Build GRCh38
rs767060558 Chr.2: 181538181-181538181 on Build GRCh38
rs770067573 Chr.2: 181538225-181538227 on Build GRCh38
rs539464631 Chr.2: 181538247-181538247 on Build GRCh38
rs535182439 Chr.2: 181538129-181538129 on Build GRCh38
rs954547032 Chr.2: 181538307-181538307 on Build GRCh38
rs760717671 Chr.2: 181538279-181538281 on Build GRCh38
rs748102604 Chr.2: 181538273-181538274 on Build GRCh38
rs987285651 Chr.2: 181538308-181538308 on Build GRCh38
rs1016582691 Chr.2: 181538260-181538260 on Build GRCh38
rs763038313 Chr.2: 181538233-181538234 on Build GRCh38
rs753367773 Chr.2: 181538204-181538204 on Build GRCh38
rs769847655 Chr.2: 181538154-181538154 on Build GRCh38
rs774260786 Chr.2: 181538162-181538162 on Build GRCh38
rs781416914 Chr.2: 181538240-181538240 on Build GRCh38
rs190730565 Chr.2: 181538127-181538127 on Build GRCh38
rs770992513 Chr.2: 181538268-181538268 on Build GRCh38
rs771621589 Chr.2: 181538186-181538187 on Build GRCh38
rs368088937 Chr.2: 181538293-181538293 on Build GRCh38
rs561713407 Chr.2: 181538127-181538156 on Build GRCh38
rs780579046 Chr.2: 181538222-181538222 on Build GRCh38
rs371788033 Chr.2: 181538249-181538249 on Build GRCh38
rs757031312 Chr.2: 181538174-181538175 on Build GRCh38
rs866647466 Chr.2: 181538251-181538251 on Build GRCh38
rs768429897 Chr.2: 181538197-181538197 on Build GRCh38
rs577490028 Chr.2: 181538242-181538242 on Build GRCh38
rs534779132 Chr.2: 181538180-181538180 on Build GRCh38
rs141723283 Chr.2: 181538227-181538227 on Build GRCh38
rs763552735 Chr.2: 181538191-181538191 on Build GRCh38
rs766312901 Chr.2: 181538135-181538153 on Build GRCh38
rs945540679 Chr.2: 181538271-181538271 on Build GRCh38
rs867507881 Chr.2: 181538215-181538215 on Build GRCh38
rs368851275 Chr.2: 181538159-181538159 on Build GRCh38
rs766477875 Chr.2: 181538160-181538160 on Build GRCh38
rs554779592 Chr.2: 181538294-181538294 on Build GRCh38
rs773497189 Chr.2: 181538228-181538229 on Build GRCh38

View Full Product Details