Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74035229-74035500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELN
Entrez Gene ID
2006
Gene Name
elastin
Gene Aliases
SVAS, WBS, WS
UniGene
Hs.647061
Gene Chromosome Location
Chr.7: 74027772-74069907 on Build GRCh38
Transcripts
NM_000501.3 NM_001081752.2 NM_001081753.2 NM_001081754.2 NM_001081755.2 NM_001278912.1 NM_001278913.1 NM_001278914.1 NM_001278915.1 NM_001278916.1 NM_001278917.1 NM_001278918.1 NM_001278939.1 XM_005250187.1 XM_005250188.1 XM_011515868.1 XM_011515869.1 XM_011515870.1 XM_011515871.1 XM_011515872.1 XM_011515873.1 XM_011515874.1 XM_011515875.1 XM_011515876.1 XM_011515877.1 XM_017011813.1 XM_017011814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640409 c.88C>T p.P30S Chr.7: 74035369-74035369 on Build GRCh38
7179683 c.130C>T p.P44S Chr.7: 74035411-74035411 on Build GRCh38
4363098 c.133+5C>T p.? Chr.7: 74035419-74035419 on Build GRCh38
3640411 c.119G>A p.G40E Chr.7: 74035400-74035400 on Build GRCh38
6871489 c.104G>A p.G35D Chr.7: 74035385-74035385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574513909 Chr.7: 74035282-74035282 on Build GRCh38
rs377303677 Chr.7: 74035452-74035452 on Build GRCh38
rs934664283 Chr.7: 74035340-74035340 on Build GRCh38
rs28763980 Chr.7: 74035425-74035425 on Build GRCh38
rs781951450 Chr.7: 74035394-74035394 on Build GRCh38
rs782707995 Chr.7: 74035429-74035430 on Build GRCh38
rs542130224 Chr.7: 74035306-74035306 on Build GRCh38
rs781876912 Chr.7: 74035408-74035408 on Build GRCh38
rs371443017 Chr.7: 74035313-74035313 on Build GRCh38
rs782193520 Chr.7: 74035365-74035365 on Build GRCh38
rs782513031 Chr.7: 74035432-74035432 on Build GRCh38
rs782625771 Chr.7: 74035373-74035373 on Build GRCh38
rs183629619 Chr.7: 74035311-74035311 on Build GRCh38
rs782808104 Chr.7: 74035421-74035421 on Build GRCh38
rs947508444 Chr.7: 74035296-74035296 on Build GRCh38
rs782647891 Chr.7: 74035332-74035332 on Build GRCh38
rs782423702 Chr.7: 74035469-74035469 on Build GRCh38
rs373683865 Chr.7: 74035346-74035346 on Build GRCh38
rs782038454 Chr.7: 74035385-74035385 on Build GRCh38
rs938974226 Chr.7: 74035254-74035254 on Build GRCh38
rs782617573 Chr.7: 74035317-74035317 on Build GRCh38
rs782104032 Chr.7: 74035395-74035395 on Build GRCh38
rs545686066 Chr.7: 74035344-74035344 on Build GRCh38
rs992132506 Chr.7: 74035274-74035274 on Build GRCh38
rs727503023 Chr.7: 74035412-74035412 on Build GRCh38
rs375144839 Chr.7: 74035419-74035419 on Build GRCh38
rs144223231 Chr.7: 74035430-74035430 on Build GRCh38
rs782391704 Chr.7: 74035376-74035376 on Build GRCh38
rs781786767 Chr.7: 74035423-74035423 on Build GRCh38
rs916116041 Chr.7: 74035281-74035281 on Build GRCh38
rs530068734 Chr.7: 74035459-74035459 on Build GRCh38
rs888291783 Chr.7: 74035464-74035464 on Build GRCh38
rs782145624 Chr.7: 74035450-74035450 on Build GRCh38
rs55951999 Chr.7: 74035400-74035400 on Build GRCh38
rs368112890 Chr.7: 74035390-74035390 on Build GRCh38
rs782517405 Chr.7: 74035330-74035330 on Build GRCh38
rs782238204 Chr.7: 74035375-74035375 on Build GRCh38
rs781844782 Chr.7: 74035322-74035322 on Build GRCh38
rs782637996 Chr.7: 74035434-74035434 on Build GRCh38
rs530703613 Chr.7: 74035341-74035341 on Build GRCh38
rs372742689 Chr.7: 74035473-74035473 on Build GRCh38

View Full Product Details