Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69256656-69256905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF6
Entrez Gene ID
11052
Gene Name
cleavage and polyadenylation specific factor 6
Gene Aliases
CFIM, CFIM68, HPBRII-4, HPBRII-7
UniGene
Hs.369606
Gene Chromosome Location
Chr.12: 69239537-69274358 on Build GRCh38
Transcripts
NM_001300947.1 NM_007007.2 XM_005268588.2 XM_005268590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998367 c.484C>A p.Q162K Chr.12: 69256806-69256806 on Build GRCh38
5754002 c.416A>G p.K139R Chr.12: 69256738-69256738 on Build GRCh38
2099619 c.430T>A p.L144I Chr.12: 69256752-69256752 on Build GRCh38
695310 c.434C>A p.P145H Chr.12: 69256756-69256756 on Build GRCh38
4210765 c.430T>A p.L144I Chr.12: 69256752-69256752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200927880 Chr.12: 69256762-69256762 on Build GRCh38
rs748145228 Chr.12: 69256871-69256871 on Build GRCh38
rs769565690 Chr.12: 69256875-69256875 on Build GRCh38
rs779896231 Chr.12: 69256785-69256785 on Build GRCh38
rs1026655411 Chr.12: 69256772-69256772 on Build GRCh38
rs995232341 Chr.12: 69256726-69256726 on Build GRCh38
rs200165673 Chr.12: 69256688-69256688 on Build GRCh38
rs986028210 Chr.12: 69256842-69256842 on Build GRCh38
rs896805021 Chr.12: 69256718-69256718 on Build GRCh38
rs369020999 Chr.12: 69256796-69256796 on Build GRCh38
rs768343669 Chr.12: 69256814-69256814 on Build GRCh38
rs753260755 Chr.12: 69256686-69256686 on Build GRCh38
rs1020933304 Chr.12: 69256759-69256759 on Build GRCh38
rs778232522 Chr.12: 69256693-69256693 on Build GRCh38
rs149215151 Chr.12: 69256751-69256751 on Build GRCh38
rs754105016 Chr.12: 69256712-69256712 on Build GRCh38
rs564462234 Chr.12: 69256864-69256864 on Build GRCh38

View Full Product Details