Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20091194-20091467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR8
Entrez Gene ID
54487
Gene Name
DGCR8 microprocessor complex subunit
Gene Aliases
C22orf12, DGCRK6, Gy1, pasha
UniGene
Hs.643452
Gene Chromosome Location
Chr.22: 20080232-20111877 on Build GRCh38
Transcripts
NM_001190326.1 NM_022720.6 XM_006724268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1682110 c.1321C>T p.R441* Chr.22: 20091449-20091449 on Build GRCh38
6642882 c.1322G>A p.R441Q Chr.22: 20091450-20091450 on Build GRCh38
4794707 c.1332G>A p.L444L Chr.22: 20091460-20091460 on Build GRCh38
1032180 c.1311C>T p.L437L Chr.22: 20091439-20091439 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765067534 Chr.22: 20091437-20091437 on Build GRCh38
rs774480219 Chr.22: 20091400-20091400 on Build GRCh38
rs376667397 Chr.22: 20091421-20091421 on Build GRCh38
rs760657180 Chr.22: 20091272-20091272 on Build GRCh38
rs993567910 Chr.22: 20091223-20091223 on Build GRCh38
rs768471115 Chr.22: 20091404-20091405 on Build GRCh38
rs977171958 Chr.22: 20091385-20091385 on Build GRCh38
rs142497183 Chr.22: 20091425-20091425 on Build GRCh38
rs558271325 Chr.22: 20091230-20091230 on Build GRCh38
rs752529619 Chr.22: 20091439-20091439 on Build GRCh38
rs1026851700 Chr.22: 20091322-20091322 on Build GRCh38
rs890715469 Chr.22: 20091250-20091250 on Build GRCh38
rs151091047 Chr.22: 20091328-20091328 on Build GRCh38
rs1017863364 Chr.22: 20091302-20091302 on Build GRCh38
rs374909151 Chr.22: 20091225-20091225 on Build GRCh38
rs762724733 Chr.22: 20091445-20091445 on Build GRCh38
rs965000607 Chr.22: 20091321-20091321 on Build GRCh38
rs578166146 Chr.22: 20091380-20091380 on Build GRCh38
rs575116026 Chr.22: 20091319-20091319 on Build GRCh38
rs538156782 Chr.22: 20091420-20091420 on Build GRCh38
rs759552286 Chr.22: 20091426-20091426 on Build GRCh38
rs372045002 Chr.22: 20091390-20091390 on Build GRCh38
rs935633142 Chr.22: 20091398-20091398 on Build GRCh38
rs973585561 Chr.22: 20091322-20091322 on Build GRCh38
rs559126853 Chr.22: 20091365-20091365 on Build GRCh38
rs1006235590 Chr.22: 20091253-20091253 on Build GRCh38
rs923495426 Chr.22: 20091346-20091346 on Build GRCh38
rs982527250 Chr.22: 20091357-20091357 on Build GRCh38

View Full Product Details