Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58096032-58096297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNB
Entrez Gene ID
2317
Gene Name
filamin B
Gene Aliases
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP
UniGene
Hs.476448
Gene Chromosome Location
Chr.3: 58008400-58172255 on Build GRCh38
Transcripts
NM_001164317.1 NM_001164318.1 NM_001164319.1 NM_001457.3 XM_005264978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5205916 c.948_949delTG p.V317fs*27 Chr.3: 58096182-58096183 on Build GRCh38
2782301 c.920_921insA p.D308fs*1 Chr.3: 58096154-58096155 on Build GRCh38
2782302 c.920_921insA p.D308fs*1 Chr.3: 58096154-58096155 on Build GRCh38
5922313 c.907-8C>T p.? Chr.3: 58096133-58096133 on Build GRCh38
5019479 c.927T>C p.S309S Chr.3: 58096161-58096161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200745639 Chr.3: 58096167-58096167 on Build GRCh38
rs934311388 Chr.3: 58096097-58096097 on Build GRCh38
rs766974304 Chr.3: 58096207-58096207 on Build GRCh38
rs923687409 Chr.3: 58096240-58096240 on Build GRCh38
rs769829306 Chr.3: 58096104-58096104 on Build GRCh38
rs754280991 Chr.3: 58096213-58096213 on Build GRCh38
rs775286882 Chr.3: 58096132-58096132 on Build GRCh38
rs989012778 Chr.3: 58096272-58096272 on Build GRCh38
rs767655909 Chr.3: 58096170-58096170 on Build GRCh38
rs191626739 Chr.3: 58096265-58096265 on Build GRCh38
rs369937858 Chr.3: 58096233-58096233 on Build GRCh38
rs776289974 Chr.3: 58096206-58096206 on Build GRCh38
rs147591946 Chr.3: 58096145-58096145 on Build GRCh38
rs750525817 Chr.3: 58096180-58096180 on Build GRCh38
rs1016019158 Chr.3: 58096126-58096126 on Build GRCh38
rs370030712 Chr.3: 58096059-58096059 on Build GRCh38
rs774083562 Chr.3: 58096160-58096160 on Build GRCh38
rs1004677351 Chr.3: 58096111-58096111 on Build GRCh38
rs991123984 Chr.3: 58096148-58096148 on Build GRCh38
rs746733392 Chr.3: 58096103-58096103 on Build GRCh38
rs762845255 Chr.3: 58096137-58096137 on Build GRCh38
rs948896228 Chr.3: 58096159-58096159 on Build GRCh38
rs1522384 Chr.3: 58096161-58096161 on Build GRCh38
rs745312071 Chr.3: 58096101-58096101 on Build GRCh38
rs755496742 Chr.3: 58096228-58096228 on Build GRCh38
rs185917667 Chr.3: 58096081-58096081 on Build GRCh38

View Full Product Details