Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158654544-158654769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM1
Entrez Gene ID
85476
Gene Name
G elongation factor mitochondrial 1
Gene Aliases
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1
UniGene
Hs.518355
Gene Chromosome Location
Chr.3: 158644497-158692572 on Build GRCh38
Transcripts
NM_001308164.1 NM_001308166.1 NM_024996.5 XM_006713795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327755 c.280C>G p.L94V Chr.3: 158654626-158654626 on Build GRCh38
3660310 c.207A>G p.S69S Chr.3: 158654553-158654553 on Build GRCh38
5814483 c.226C>A p.L76I Chr.3: 158654572-158654572 on Build GRCh38
3940367 c.210A>G p.K70K Chr.3: 158654556-158654556 on Build GRCh38
7239436 c.1052A>G p.H351R Chr.3: 158654600-158654600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142919829 Chr.3: 158654637-158654637 on Build GRCh38
rs780884783 Chr.3: 158654683-158654683 on Build GRCh38
rs762640437 Chr.3: 158654600-158654600 on Build GRCh38
rs373952002 Chr.3: 158654580-158654580 on Build GRCh38
rs746788195 Chr.3: 158654740-158654740 on Build GRCh38
rs62286656 Chr.3: 158654688-158654688 on Build GRCh38
rs774849214 Chr.3: 158654608-158654608 on Build GRCh38
rs768006278 Chr.3: 158654632-158654632 on Build GRCh38
rs374592703 Chr.3: 158654717-158654717 on Build GRCh38
rs748044303 Chr.3: 158654593-158654593 on Build GRCh38
rs541525012 Chr.3: 158654714-158654714 on Build GRCh38
rs754703720 Chr.3: 158654672-158654672 on Build GRCh38
rs940711263 Chr.3: 158654729-158654729 on Build GRCh38
rs757468938 Chr.3: 158654642-158654642 on Build GRCh38
rs766172439 Chr.3: 158654604-158654604 on Build GRCh38
rs768570649 Chr.3: 158654575-158654575 on Build GRCh38
rs377068967 Chr.3: 158654626-158654626 on Build GRCh38
rs1040378688 Chr.3: 158654591-158654591 on Build GRCh38
rs753905803 Chr.3: 158654639-158654639 on Build GRCh38
rs187690169 Chr.3: 158654634-158654634 on Build GRCh38
rs1020182837 Chr.3: 158654694-158654694 on Build GRCh38
rs779166142 Chr.3: 158654662-158654662 on Build GRCh38
rs386667496 Chr.3: 158654688-158654713 on Build GRCh38
rs9884039 Chr.3: 158654713-158654713 on Build GRCh38
rs541257372 Chr.3: 158654649-158654649 on Build GRCh38
rs769311074 Chr.3: 158654597-158654597 on Build GRCh38
rs1019230632 Chr.3: 158654594-158654594 on Build GRCh38
rs146827771 Chr.3: 158654573-158654573 on Build GRCh38

View Full Product Details