Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64117850-64118079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
UniGene
Hs.584876
Gene Chromosome Location
Chr.11: 64035970-64119183 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_011544970.2

Gene Symbol
MACROD1
Entrez Gene ID
28992
Gene Name
MACRO domain containing 1
Gene Aliases
LRP16
UniGene
Hs.602898
Gene Chromosome Location
Chr.11: 63998554-64166652 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_011544970.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665167 c.1633G>A p.A545T Chr.11: 64117900-64117900 on Build GRCh38
7261326 c.1715C>G p.A572G Chr.11: 64117982-64117982 on Build GRCh38
5811753 c.1674C>T p.G558G Chr.11: 64117941-64117941 on Build GRCh38
1661874 c.1705G>A p.V569I Chr.11: 64117972-64117972 on Build GRCh38
7207197 c.1763G>C p.R588T Chr.11: 64118030-64118030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780913370 Chr.11: 64117972-64117972 on Build GRCh38
rs763526629 Chr.11: 64117921-64117921 on Build GRCh38
rs749335298 Chr.11: 64117994-64117994 on Build GRCh38
rs754446142 Chr.11: 64117883-64117883 on Build GRCh38
rs375487485 Chr.11: 64118041-64118041 on Build GRCh38
rs750148970 Chr.11: 64118045-64118045 on Build GRCh38
rs770731645 Chr.11: 64117902-64117902 on Build GRCh38
rs751508281 Chr.11: 64117969-64117969 on Build GRCh38
rs764755729 Chr.11: 64118011-64118011 on Build GRCh38
rs749527475 Chr.11: 64118051-64118051 on Build GRCh38
rs762387650 Chr.11: 64118024-64118024 on Build GRCh38
rs778352526 Chr.11: 64117896-64117896 on Build GRCh38
rs140274429 Chr.11: 64117996-64117996 on Build GRCh38
rs554837203 Chr.11: 64117971-64117971 on Build GRCh38
rs148484621 Chr.11: 64117913-64117913 on Build GRCh38
rs781414347 Chr.11: 64117906-64117906 on Build GRCh38
rs201043470 Chr.11: 64117962-64117962 on Build GRCh38
rs566001505 Chr.11: 64117952-64117952 on Build GRCh38
rs752665635 Chr.11: 64117898-64117898 on Build GRCh38
rs200134025 Chr.11: 64118010-64118010 on Build GRCh38
rs769250709 Chr.11: 64117929-64117929 on Build GRCh38
rs142685980 Chr.11: 64117899-64117899 on Build GRCh38
rs773496799 Chr.11: 64118004-64118004 on Build GRCh38
rs372121350 Chr.11: 64117986-64117986 on Build GRCh38
rs763595179 Chr.11: 64118027-64118027 on Build GRCh38
rs769906902 Chr.11: 64117917-64117917 on Build GRCh38
rs753447977 Chr.11: 64117878-64117878 on Build GRCh38
rs143309484 Chr.11: 64117979-64117979 on Build GRCh38
rs978450483 Chr.11: 64118002-64118002 on Build GRCh38
rs142645704 Chr.11: 64117928-64117928 on Build GRCh38
rs533249444 Chr.11: 64118046-64118046 on Build GRCh38
rs150943813 Chr.11: 64117939-64117939 on Build GRCh38
rs760989622 Chr.11: 64117942-64117942 on Build GRCh38
rs767315617 Chr.11: 64118042-64118042 on Build GRCh38
rs757935460 Chr.11: 64117956-64117956 on Build GRCh38
rs746762287 Chr.11: 64117900-64117900 on Build GRCh38
rs140790562 Chr.11: 64117959-64117959 on Build GRCh38
rs373807742 Chr.11: 64117920-64117920 on Build GRCh38
rs911603132 Chr.11: 64117876-64117876 on Build GRCh38
rs200955257 Chr.11: 64117938-64117938 on Build GRCh38
rs746082742 Chr.11: 64117977-64117977 on Build GRCh38
rs150249733 Chr.11: 64118047-64118047 on Build GRCh38
rs764780926 Chr.11: 64117951-64117951 on Build GRCh38
rs766081520 Chr.11: 64117869-64117869 on Build GRCh38
rs768000188 Chr.11: 64117941-64117941 on Build GRCh38
rs35851449 Chr.11: 64117995-64117995 on Build GRCh38
rs555915933 Chr.11: 64118023-64118023 on Build GRCh38

View Full Product Details