Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30595111-30595372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GARS
Entrez Gene ID
2617
Gene Name
glycyl-tRNA synthetase
Gene Aliases
CMT2D, DSMAV, GlyRS, HMN5, SMAD1
UniGene
Hs.404321
Gene Chromosome Location
Chr.7: 30594735-30634033 on Build GRCh38
Transcripts
NM_001316772.1 NM_002047.3 XM_006715686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763781 c.218A>G p.Q73R Chr.7: 30595139-30595139 on Build GRCh38
6395082 c.193G>A p.A65T Chr.7: 30595114-30595114 on Build GRCh38
5764896 c.222+5C>T p.? Chr.7: 30595148-30595148 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763562755 Chr.7: 30595158-30595158 on Build GRCh38
rs1006993591 Chr.7: 30595226-30595226 on Build GRCh38
rs564113357 Chr.7: 30595308-30595308 on Build GRCh38
rs542478317 Chr.7: 30595219-30595219 on Build GRCh38
rs778425012 Chr.7: 30595274-30595274 on Build GRCh38
rs188159858 Chr.7: 30595292-30595292 on Build GRCh38
rs755747869 Chr.7: 30595196-30595196 on Build GRCh38
rs1001683268 Chr.7: 30595323-30595323 on Build GRCh38
rs980682461 Chr.7: 30595186-30595186 on Build GRCh38
rs906135358 Chr.7: 30595310-30595310 on Build GRCh38
rs1035413546 Chr.7: 30595189-30595189 on Build GRCh38
rs556701108 Chr.7: 30595131-30595131 on Build GRCh38
rs1052045624 Chr.7: 30595237-30595237 on Build GRCh38
rs947065640 Chr.7: 30595216-30595216 on Build GRCh38
rs921884865 Chr.7: 30595232-30595234 on Build GRCh38
rs891187373 Chr.7: 30595263-30595263 on Build GRCh38
rs1019604005 Chr.7: 30595247-30595247 on Build GRCh38
rs752272200 Chr.7: 30595195-30595195 on Build GRCh38
rs2072236 Chr.7: 30595148-30595148 on Build GRCh38
rs965394954 Chr.7: 30595266-30595266 on Build GRCh38
rs769658988 Chr.7: 30595137-30595137 on Build GRCh38
rs924559604 Chr.7: 30595278-30595278 on Build GRCh38
rs931768382 Chr.7: 30595236-30595236 on Build GRCh38
rs771148911 Chr.7: 30595313-30595313 on Build GRCh38
rs1008199275 Chr.7: 30595129-30595129 on Build GRCh38
rs761512824 Chr.7: 30595176-30595176 on Build GRCh38
rs766720408 Chr.7: 30595181-30595181 on Build GRCh38
rs773923827 Chr.7: 30595166-30595166 on Build GRCh38
rs553636445 Chr.7: 30595194-30595196 on Build GRCh38
rs990351892 Chr.7: 30595290-30595290 on Build GRCh38
rs1019128382 Chr.7: 30595276-30595277 on Build GRCh38
rs775001026 Chr.7: 30595138-30595138 on Build GRCh38
rs759393844 Chr.7: 30595136-30595136 on Build GRCh38
rs538855762 Chr.7: 30595284-30595284 on Build GRCh38
rs554181658 Chr.7: 30595269-30595269 on Build GRCh38
rs1027068048 Chr.7: 30595208-30595208 on Build GRCh38

View Full Product Details