Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42728079-42728310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670586 c.2868+1G>A p.? Chr.15: 42728203-42728203 on Build GRCh38
5422806 c.2849C>T p.P950L Chr.15: 42728223-42728223 on Build GRCh38
6346472 c.2836C>A p.R946R Chr.15: 42728236-42728236 on Build GRCh38
2187687 c.2862G>A p.P954P Chr.15: 42728210-42728210 on Build GRCh38
7360343 c.2837G>A p.R946Q Chr.15: 42728235-42728235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556046335 Chr.15: 42728099-42728099 on Build GRCh38
rs754625040 Chr.15: 42728159-42728159 on Build GRCh38
rs756751239 Chr.15: 42728183-42728183 on Build GRCh38
rs777603482 Chr.15: 42728159-42728161 on Build GRCh38
rs758691277 Chr.15: 42728276-42728276 on Build GRCh38
rs1041106624 Chr.15: 42728219-42728219 on Build GRCh38
rs754334399 Chr.15: 42728243-42728243 on Build GRCh38
rs768041338 Chr.15: 42728171-42728171 on Build GRCh38
rs745898297 Chr.15: 42728167-42728186 on Build GRCh38
rs145148854 Chr.15: 42728203-42728203 on Build GRCh38
rs150438471 Chr.15: 42728204-42728204 on Build GRCh38
rs778130045 Chr.15: 42728186-42728186 on Build GRCh38
rs753079849 Chr.15: 42728182-42728182 on Build GRCh38
rs146535176 Chr.15: 42728115-42728115 on Build GRCh38
rs931460177 Chr.15: 42728163-42728163 on Build GRCh38
rs200767055 Chr.15: 42728278-42728278 on Build GRCh38
rs1039455419 Chr.15: 42728196-42728196 on Build GRCh38
rs550681477 Chr.15: 42728123-42728123 on Build GRCh38
rs377606247 Chr.15: 42728149-42728149 on Build GRCh38
rs773023944 Chr.15: 42728211-42728211 on Build GRCh38
rs541034913 Chr.15: 42728112-42728112 on Build GRCh38
rs747935657 Chr.15: 42728210-42728210 on Build GRCh38
rs138228278 Chr.15: 42728231-42728231 on Build GRCh38
rs751198894 Chr.15: 42728158-42728158 on Build GRCh38
rs142833783 Chr.15: 42728235-42728235 on Build GRCh38
rs139032045 Chr.15: 42728270-42728270 on Build GRCh38
rs201757681 Chr.15: 42728232-42728232 on Build GRCh38
rs546806428 Chr.15: 42728147-42728147 on Build GRCh38
rs62020665 Chr.15: 42728126-42728126 on Build GRCh38
rs759300971 Chr.15: 42728233-42728233 on Build GRCh38
rs749746339 Chr.15: 42728200-42728200 on Build GRCh38
rs141236475 Chr.15: 42728130-42728130 on Build GRCh38
rs559228588 Chr.15: 42728264-42728264 on Build GRCh38
rs1048559811 Chr.15: 42728197-42728197 on Build GRCh38
rs114779238 Chr.15: 42728236-42728236 on Build GRCh38
rs117597169 Chr.15: 42728100-42728100 on Build GRCh38
rs375929824 Chr.15: 42728285-42728285 on Build GRCh38
rs916471417 Chr.15: 42728120-42728120 on Build GRCh38
rs1026619957 Chr.15: 42728209-42728209 on Build GRCh38
rs769503711 Chr.15: 42728284-42728284 on Build GRCh38
rs772262599 Chr.15: 42728205-42728205 on Build GRCh38
rs747164189 Chr.15: 42728282-42728282 on Build GRCh38
rs776454812 Chr.15: 42728207-42728207 on Build GRCh38
rs938755549 Chr.15: 42728131-42728131 on Build GRCh38
rs757734894 Chr.15: 42728245-42728245 on Build GRCh38
rs1052326712 Chr.15: 42728148-42728148 on Build GRCh38
rs1037087427 Chr.15: 42728143-42728143 on Build GRCh38
rs943904295 Chr.15: 42728167-42728167 on Build GRCh38
rs762904904 Chr.15: 42728223-42728223 on Build GRCh38
rs756449578 Chr.15: 42728239-42728239 on Build GRCh38

View Full Product Details