Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6845725-6845972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA3
Entrez Gene ID
83461
Gene Name
cell division cycle associated 3
Gene Aliases
GRCC8, TOME-1
UniGene
Hs.524216
Gene Chromosome Location
Chr.12: 6844796-6852066 on Build GRCh38
Transcripts
NM_001297571.1 NM_002075.3 XM_011520953.2

Gene Symbol
GNB3
Entrez Gene ID
2784
Gene Name
G protein subunit beta 3
Gene Aliases
CSNB1H
UniGene
Hs.631657
Gene Chromosome Location
Chr.12: 6840854-6847400 on Build GRCh38
Transcripts
NM_001297571.1 NM_002075.3 XM_011520953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311026 c.911G>T p.R304L Chr.12: 6845797-6845797 on Build GRCh38
1363933 c.848G>A p.R283H Chr.12: 6845734-6845734 on Build GRCh38
1363936 c.910C>T p.R304C Chr.12: 6845796-6845796 on Build GRCh38
5519112 c.916+7G>A p.? Chr.12: 6845809-6845809 on Build GRCh38
695313 c.873C>T p.D291D Chr.12: 6845759-6845759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895627196 Chr.12: 6845750-6845750 on Build GRCh38
rs782626186 Chr.12: 6845796-6845796 on Build GRCh38
rs200733665 Chr.12: 6845766-6845766 on Build GRCh38
rs782275874 Chr.12: 6845786-6845786 on Build GRCh38
rs782065330 Chr.12: 6845836-6845836 on Build GRCh38
rs782729041 Chr.12: 6845757-6845757 on Build GRCh38
rs376237808 Chr.12: 6845809-6845809 on Build GRCh38
rs782090563 Chr.12: 6845745-6845745 on Build GRCh38
rs140337095 Chr.12: 6845780-6845780 on Build GRCh38
rs782531925 Chr.12: 6845787-6845787 on Build GRCh38
rs780628128 Chr.12: 6845823-6845823 on Build GRCh38
rs781848235 Chr.12: 6845755-6845755 on Build GRCh38
rs201032638 Chr.12: 6845829-6845829 on Build GRCh38
rs535223865 Chr.12: 6845881-6845881 on Build GRCh38
rs782189035 Chr.12: 6845803-6845803 on Build GRCh38
rs781818020 Chr.12: 6845874-6845874 on Build GRCh38
rs543958987 Chr.12: 6845935-6845935 on Build GRCh38
rs782763999 Chr.12: 6845754-6845754 on Build GRCh38
rs782139192 Chr.12: 6845753-6845753 on Build GRCh38
rs782760944 Chr.12: 6845833-6845833 on Build GRCh38
rs782692758 Chr.12: 6845781-6845781 on Build GRCh38
rs782087746 Chr.12: 6845844-6845844 on Build GRCh38
rs1017383822 Chr.12: 6845936-6845936 on Build GRCh38
rs782103541 Chr.12: 6845830-6845830 on Build GRCh38
rs753136154 Chr.12: 6845756-6845756 on Build GRCh38
rs376517711 Chr.12: 6845744-6845744 on Build GRCh38
rs144342814 Chr.12: 6845797-6845797 on Build GRCh38
rs782760885 Chr.12: 6845835-6845835 on Build GRCh38
rs782401589 Chr.12: 6845895-6845895 on Build GRCh38
rs1044470016 Chr.12: 6845894-6845894 on Build GRCh38
rs949898567 Chr.12: 6845794-6845794 on Build GRCh38
rs781804029 Chr.12: 6845777-6845777 on Build GRCh38

View Full Product Details