Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66940496-66940759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES2
Entrez Gene ID
8824
Gene Name
carboxylesterase 2
Gene Aliases
CE-2, CES2A1, PCE-2, iCE
UniGene
Hs.282975
Gene Chromosome Location
Chr.16: 66934444-66945096 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 XM_011523421.2 XM_017023818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1167671 c.879delG p.G294fs*80 Chr.16: 66940566-66940566 on Build GRCh38
3510875 c.911C>T p.S304F Chr.16: 66940598-66940598 on Build GRCh38
7162944 c.945C>T p.I315I Chr.16: 66940632-66940632 on Build GRCh38
5851189 c.838G>T p.G280* Chr.16: 66940525-66940525 on Build GRCh38
3969634 c.888G>A p.T296T Chr.16: 66940575-66940575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769906099 Chr.16: 66940634-66940634 on Build GRCh38
rs770907161 Chr.16: 66940728-66940728 on Build GRCh38
rs780237910 Chr.16: 66940643-66940643 on Build GRCh38
rs745968666 Chr.16: 66940560-66940560 on Build GRCh38
rs748496522 Chr.16: 66940541-66940541 on Build GRCh38
rs780784233 Chr.16: 66940674-66940674 on Build GRCh38
rs747620099 Chr.16: 66940554-66940554 on Build GRCh38
rs1011400116 Chr.16: 66940583-66940583 on Build GRCh38
rs112732183 Chr.16: 66940516-66940516 on Build GRCh38
rs758846014 Chr.16: 66940542-66940542 on Build GRCh38
rs778134567 Chr.16: 66940620-66940620 on Build GRCh38
rs532312387 Chr.16: 66940517-66940517 on Build GRCh38
rs764961386 Chr.16: 66940591-66940591 on Build GRCh38
rs150055059 Chr.16: 66940566-66940566 on Build GRCh38
rs781686511 Chr.16: 66940628-66940628 on Build GRCh38
rs781645148 Chr.16: 66940685-66940685 on Build GRCh38
rs757657844 Chr.16: 66940684-66940684 on Build GRCh38
rs145407778 Chr.16: 66940586-66940586 on Build GRCh38
rs769964375 Chr.16: 66940561-66940561 on Build GRCh38
rs142398307 Chr.16: 66940632-66940632 on Build GRCh38
rs773014158 Chr.16: 66940648-66940648 on Build GRCh38
rs746800741 Chr.16: 66940724-66940724 on Build GRCh38
rs867584938 Chr.16: 66940535-66940535 on Build GRCh38
rs749351541 Chr.16: 66940703-66940703 on Build GRCh38
rs752992041 Chr.16: 66940703-66940703 on Build GRCh38
rs780112513 Chr.16: 66940699-66940699 on Build GRCh38
rs141625476 Chr.16: 66940694-66940694 on Build GRCh38
rs768763536 Chr.16: 66940645-66940645 on Build GRCh38
rs372139651 Chr.16: 66940660-66940660 on Build GRCh38
rs374880110 Chr.16: 66940672-66940672 on Build GRCh38
rs149179989 Chr.16: 66940587-66940587 on Build GRCh38
rs10852434 Chr.16: 66940624-66940624 on Build GRCh38
rs552023858 Chr.16: 66940540-66940540 on Build GRCh38
rs763673959 Chr.16: 66940678-66940678 on Build GRCh38
rs182454684 Chr.16: 66940578-66940578 on Build GRCh38
rs753414364 Chr.16: 66940532-66940532 on Build GRCh38
rs142953579 Chr.16: 66940695-66940695 on Build GRCh38
rs139112900 Chr.16: 66940539-66940539 on Build GRCh38
rs778129977 Chr.16: 66940543-66940543 on Build GRCh38
rs768033054 Chr.16: 66940538-66940545 on Build GRCh38
rs371064921 Chr.16: 66940565-66940565 on Build GRCh38
rs201663206 Chr.16: 66940558-66940558 on Build GRCh38
rs375373068 Chr.16: 66940623-66940623 on Build GRCh38
rs768885998 Chr.16: 66940705-66940705 on Build GRCh38
rs752154344 Chr.16: 66940671-66940671 on Build GRCh38
rs773015939 Chr.16: 66940573-66940573 on Build GRCh38
rs967462325 Chr.16: 66940736-66940736 on Build GRCh38
rs779047656 Chr.16: 66940712-66940712 on Build GRCh38
rs770881172 Chr.16: 66940665-66940665 on Build GRCh38
rs147942040 Chr.16: 66940657-66940657 on Build GRCh38
rs759103977 Chr.16: 66940668-66940668 on Build GRCh38
rs954420905 Chr.16: 66940625-66940625 on Build GRCh38
rs764908323 Chr.16: 66940670-66940670 on Build GRCh38
rs758686915 Chr.16: 66940603-66940603 on Build GRCh38
rs1020316023 Chr.16: 66940730-66940730 on Build GRCh38
rs1052988153 Chr.16: 66940569-66940569 on Build GRCh38
rs534369649 Chr.16: 66940574-66940574 on Build GRCh38
rs200667205 Chr.16: 66940557-66940557 on Build GRCh38
rs770964350 Chr.16: 66940659-66940659 on Build GRCh38
rs556416386 Chr.16: 66940644-66940644 on Build GRCh38
rs752453143 Chr.16: 66940596-66940596 on Build GRCh38

View Full Product Details