Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 1053629-1053898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
UniGene
Hs.59506
Gene Chromosome Location
Chr.9: 1003237-1057554 on Build GRCh38
Transcripts
NM_001130865.2 NM_006557.6 NM_181872.4 XM_011517687.1 XM_011517690.2 XM_011517694.2 XM_017014213.1 XM_017014214.1 XM_017014215.1 XM_017014216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6182187 c.555T>A p.N185K Chr.9: 1053751-1053751 on Build GRCh38
6182185 c.566G>A p.R189H Chr.9: 1053762-1053762 on Build GRCh38
5557734 c.534G>A p.K178K Chr.9: 1053730-1053730 on Build GRCh38
1726613 c.595G>T p.A199S Chr.9: 1053791-1053791 on Build GRCh38
1250268 c.560T>A p.F187Y Chr.9: 1053756-1053756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762653085 Chr.9: 1053831-1053831 on Build GRCh38
rs561965987 Chr.9: 1053834-1053834 on Build GRCh38
rs770503994 Chr.9: 1053722-1053722 on Build GRCh38
rs939284951 Chr.9: 1053657-1053657 on Build GRCh38
rs200011939 Chr.9: 1053762-1053762 on Build GRCh38
rs895516864 Chr.9: 1053752-1053752 on Build GRCh38
rs779382151 Chr.9: 1053785-1053785 on Build GRCh38
rs778239045 Chr.9: 1053856-1053856 on Build GRCh38
rs776244117 Chr.9: 1053808-1053808 on Build GRCh38
rs775413150 Chr.9: 1053820-1053820 on Build GRCh38
rs772698447 Chr.9: 1053797-1053797 on Build GRCh38
rs772911244 Chr.9: 1053713-1053713 on Build GRCh38
rs779104934 Chr.9: 1053691-1053691 on Build GRCh38
rs759064574 Chr.9: 1053734-1053734 on Build GRCh38
rs776439646 Chr.9: 1053724-1053724 on Build GRCh38
rs748727759 Chr.9: 1053704-1053704 on Build GRCh38
rs781279164 Chr.9: 1053767-1053767 on Build GRCh38
rs770591982 Chr.9: 1053803-1053803 on Build GRCh38
rs945213517 Chr.9: 1053700-1053700 on Build GRCh38
rs778425270 Chr.9: 1053798-1053798 on Build GRCh38
rs760253944 Chr.9: 1053717-1053717 on Build GRCh38
rs144715662 Chr.9: 1053765-1053765 on Build GRCh38
rs759276825 Chr.9: 1053813-1053813 on Build GRCh38
rs769311515 Chr.9: 1053819-1053819 on Build GRCh38
rs754089755 Chr.9: 1053772-1053772 on Build GRCh38
rs751390405 Chr.9: 1053671-1053671 on Build GRCh38
rs769086115 Chr.9: 1053689-1053689 on Build GRCh38
rs752405627 Chr.9: 1053748-1053748 on Build GRCh38
rs559419614 Chr.9: 1053739-1053739 on Build GRCh38
rs528392769 Chr.9: 1053747-1053747 on Build GRCh38
rs749452787 Chr.9: 1053678-1053678 on Build GRCh38
rs776402733 Chr.9: 1053666-1053666 on Build GRCh38
rs752894563 Chr.9: 1053849-1053849 on Build GRCh38
rs757811398 Chr.9: 1053863-1053863 on Build GRCh38
rs778994545 Chr.9: 1053847-1053847 on Build GRCh38
rs747552324 Chr.9: 1053861-1053861 on Build GRCh38
rs753836628 Chr.9: 1053843-1053843 on Build GRCh38
rs766484682 Chr.9: 1053841-1053841 on Build GRCh38
rs2275517 Chr.9: 1053665-1053665 on Build GRCh38
rs147349546 Chr.9: 1053656-1053656 on Build GRCh38
rs769556450 Chr.9: 1053867-1053867 on Build GRCh38
rs201585505 Chr.9: 1053761-1053761 on Build GRCh38
rs926523178 Chr.9: 1053655-1053655 on Build GRCh38
rs745359257 Chr.9: 1053866-1053866 on Build GRCh38
rs772432457 Chr.9: 1053705-1053705 on Build GRCh38
rs747464257 Chr.9: 1053802-1053802 on Build GRCh38
rs750568280 Chr.9: 1053763-1053763 on Build GRCh38
rs755122224 Chr.9: 1053774-1053774 on Build GRCh38
rs991975279 Chr.9: 1053664-1053664 on Build GRCh38
rs758707176 Chr.9: 1053855-1053855 on Build GRCh38

View Full Product Details