Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59323043-59323282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM1
Entrez Gene ID
23002
Gene Name
dishevelled associated activator of morphogenesis 1
Gene Aliases
-
UniGene
Hs.19156
Gene Chromosome Location
Chr.14: 59188611-59371405 on Build GRCh38
Transcripts
NM_001270520.1 NM_014992.2 XM_005267430.1 XM_005267431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3754020 c.723G>T p.L241L Chr.14: 59323174-59323174 on Build GRCh38
1477651 c.645G>C p.E215D Chr.14: 59323096-59323096 on Build GRCh38
1578858 c.657G>A p.T219T Chr.14: 59323108-59323108 on Build GRCh38
3814996 c.772C>G p.Q258E Chr.14: 59323223-59323223 on Build GRCh38
6042786 c.702C>T p.P234P Chr.14: 59323153-59323153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564090602 Chr.14: 59323139-59323139 on Build GRCh38
rs902156580 Chr.14: 59323193-59323193 on Build GRCh38
rs763917911 Chr.14: 59323216-59323216 on Build GRCh38
rs757156688 Chr.14: 59323150-59323150 on Build GRCh38
rs778249260 Chr.14: 59323168-59323168 on Build GRCh38
rs773808956 Chr.14: 59323098-59323098 on Build GRCh38
rs749844960 Chr.14: 59323153-59323153 on Build GRCh38
rs760420558 Chr.14: 59323135-59323135 on Build GRCh38
rs758073917 Chr.14: 59323156-59323156 on Build GRCh38
rs748395859 Chr.14: 59323077-59323077 on Build GRCh38
rs748543467 Chr.14: 59323164-59323164 on Build GRCh38
rs377002769 Chr.14: 59323117-59323117 on Build GRCh38
rs781630768 Chr.14: 59323076-59323076 on Build GRCh38
rs770457227 Chr.14: 59323093-59323093 on Build GRCh38
rs141883625 Chr.14: 59323207-59323207 on Build GRCh38
rs776405966 Chr.14: 59323221-59323221 on Build GRCh38
rs531992927 Chr.14: 59323180-59323180 on Build GRCh38
rs749794234 Chr.14: 59323102-59323102 on Build GRCh38
rs17096074 Chr.14: 59323174-59323174 on Build GRCh38
rs980120198 Chr.14: 59323172-59323172 on Build GRCh38
rs531300580 Chr.14: 59323138-59323138 on Build GRCh38
rs895420481 Chr.14: 59323228-59323228 on Build GRCh38
rs534551526 Chr.14: 59323255-59323255 on Build GRCh38
rs567041153 Chr.14: 59323208-59323208 on Build GRCh38
rs756593611 Chr.14: 59323167-59323167 on Build GRCh38
rs761753130 Chr.14: 59323230-59323230 on Build GRCh38
rs201077027 Chr.14: 59323160-59323160 on Build GRCh38
rs111578221 Chr.14: 59323236-59323236 on Build GRCh38
rs771178796 Chr.14: 59323111-59323111 on Build GRCh38
rs879260041 Chr.14: 59323107-59323107 on Build GRCh38
rs969334442 Chr.14: 59323140-59323140 on Build GRCh38
rs771717982 Chr.14: 59323233-59323233 on Build GRCh38
rs759856885 Chr.14: 59323118-59323118 on Build GRCh38
rs767573792 Chr.14: 59323120-59323120 on Build GRCh38
rs749978966 Chr.14: 59323242-59323242 on Build GRCh38
rs946478824 Chr.14: 59323154-59323154 on Build GRCh38
rs542286199 Chr.14: 59323253-59323253 on Build GRCh38
rs774506910 Chr.14: 59323194-59323194 on Build GRCh38
rs1003237514 Chr.14: 59323200-59323200 on Build GRCh38
rs775357562 Chr.14: 59323130-59323130 on Build GRCh38
rs866648375 Chr.14: 59323146-59323146 on Build GRCh38

View Full Product Details