Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 532274-532535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
UniGene
Hs.484412
Gene Chromosome Location
Chr.6: 485138-693141 on Build GRCh38
Transcripts
NM_018303.5 NR_073064.1 XM_017011018.1 XM_017011019.1 XM_017011020.1 XM_017011021.1 XM_017011022.1 XM_017011023.1 XM_017011024.1 XM_017011025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1444940 c.2349T>G p.Y783* Chr.6: 532500-532500 on Build GRCh38
94259 c.2318G>T p.G773V Chr.6: 532531-532531 on Build GRCh38
6880880 c.2377A>G p.T793A Chr.6: 532472-532472 on Build GRCh38
1734696 c.2314G>A p.V772I Chr.6: 532535-532535 on Build GRCh38
4977256 c.2334A>G p.G778G Chr.6: 532515-532515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955955701 Chr.6: 532362-532362 on Build GRCh38
rs375110429 Chr.6: 532461-532461 on Build GRCh38
rs757236060 Chr.6: 532451-532451 on Build GRCh38
rs374794273 Chr.6: 532440-532440 on Build GRCh38
rs1033353543 Chr.6: 532362-532363 on Build GRCh38
rs774963703 Chr.6: 532434-532434 on Build GRCh38
rs761785633 Chr.6: 532356-532356 on Build GRCh38
rs1000904664 Chr.6: 532413-532413 on Build GRCh38
rs776085771 Chr.6: 532487-532487 on Build GRCh38
rs760092658 Chr.6: 532439-532439 on Build GRCh38
rs371459445 Chr.6: 532376-532376 on Build GRCh38
rs201407807 Chr.6: 532486-532486 on Build GRCh38
rs982990686 Chr.6: 532464-532464 on Build GRCh38
rs1009416119 Chr.6: 532358-532358 on Build GRCh38
rs1021100285 Chr.6: 532397-532397 on Build GRCh38
rs754104808 Chr.6: 532449-532449 on Build GRCh38
rs767586968 Chr.6: 532442-532442 on Build GRCh38
rs908583656 Chr.6: 532465-532465 on Build GRCh38
rs1033341736 Chr.6: 532424-532424 on Build GRCh38
rs747869211 Chr.6: 532510-532510 on Build GRCh38
rs199870373 Chr.6: 532454-532454 on Build GRCh38
rs890945914 Chr.6: 532332-532332 on Build GRCh38
rs577483613 Chr.6: 532411-532411 on Build GRCh38
rs991680689 Chr.6: 532441-532441 on Build GRCh38
rs369805517 Chr.6: 532472-532472 on Build GRCh38
rs778995939 Chr.6: 532456-532456 on Build GRCh38
rs145082033 Chr.6: 532443-532443 on Build GRCh38
rs746772895 Chr.6: 532477-532477 on Build GRCh38
rs77649909 Chr.6: 532445-532445 on Build GRCh38

View Full Product Details