Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41683634-41683885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM7
Entrez Gene ID
1087
Gene Name
carcinoembryonic antigen related cell adhesion molecule 7
Gene Aliases
CGM2
UniGene
Hs.74466
Gene Chromosome Location
Chr.19: 41673307-41688277 on Build GRCh38
Transcripts
NM_001291485.1 NM_006890.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275537 c.686C>A p.P229Q Chr.19: 41683805-41683805 on Build GRCh38
3534450 c.680G>A p.S227N Chr.19: 41683811-41683811 on Build GRCh38
2151389 c.676C>T p.R226C Chr.19: 41683815-41683815 on Build GRCh38
4078555 c.615C>T p.S205S Chr.19: 41683876-41683876 on Build GRCh38
3892665 c.649G>A p.E217K Chr.19: 41683842-41683842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782336487 Chr.19: 41683784-41683784 on Build GRCh38
rs781783933 Chr.19: 41683852-41683852 on Build GRCh38
rs935419319 Chr.19: 41683801-41683801 on Build GRCh38
rs782087281 Chr.19: 41683819-41683819 on Build GRCh38
rs781948927 Chr.19: 41683718-41683718 on Build GRCh38
rs111534806 Chr.19: 41683788-41683788 on Build GRCh38
rs781864443 Chr.19: 41683831-41683831 on Build GRCh38
rs782558460 Chr.19: 41683740-41683740 on Build GRCh38
rs375263313 Chr.19: 41683797-41683797 on Build GRCh38
rs369677539 Chr.19: 41683815-41683815 on Build GRCh38
rs782807535 Chr.19: 41683850-41683850 on Build GRCh38
rs376404757 Chr.19: 41683859-41683859 on Build GRCh38
rs16975478 Chr.19: 41683785-41683785 on Build GRCh38
rs782187498 Chr.19: 41683762-41683762 on Build GRCh38
rs782146934 Chr.19: 41683789-41683789 on Build GRCh38
rs928142700 Chr.19: 41683720-41683720 on Build GRCh38
rs781884943 Chr.19: 41683739-41683739 on Build GRCh38
rs782553889 Chr.19: 41683755-41683755 on Build GRCh38
rs369291070 Chr.19: 41683738-41683738 on Build GRCh38
rs782307166 Chr.19: 41683814-41683814 on Build GRCh38
rs782509312 Chr.19: 41683863-41683863 on Build GRCh38
rs782440343 Chr.19: 41683857-41683857 on Build GRCh38
rs373169617 Chr.19: 41683820-41683820 on Build GRCh38

View Full Product Details