Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5821667-5821940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRMP1
Entrez Gene ID
1400
Gene Name
collapsin response mediator protein 1
Gene Aliases
CRMP-1, DPYSL1, DRP-1, DRP1, ULIP-3
UniGene
Hs.135270
Gene Chromosome Location
Chr.4: 5820764-5893083 on Build GRCh38
Transcripts
NM_001014809.2 NM_001288661.1 NM_001288662.1 NM_001313.4

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
UniGene
Hs.646899
Gene Chromosome Location
Chr.4: 5711197-5829043 on Build GRCh38
Transcripts
NM_001014809.2 NM_001288661.1 NM_001288662.1 NM_001313.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5700223 c.2016C>A p.I672I Chr.4: 5821805-5821805 on Build GRCh38
5431139 c.2009A>G p.H670R Chr.4: 5821812-5821812 on Build GRCh38
5700224 c.1674C>A p.I558I Chr.4: 5821805-5821805 on Build GRCh38
4632366 c.2005G>A p.G669S Chr.4: 5821816-5821816 on Build GRCh38
5891793 c.1675G>C p.V559L Chr.4: 5821804-5821804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191530433 Chr.4: 5821892-5821892 on Build GRCh38
rs989696923 Chr.4: 5821878-5821878 on Build GRCh38
rs776901530 Chr.4: 5821771-5821771 on Build GRCh38
rs543989909 Chr.4: 5821790-5821790 on Build GRCh38
rs747541075 Chr.4: 5821901-5821901 on Build GRCh38
rs368527112 Chr.4: 5821863-5821863 on Build GRCh38
rs749553069 Chr.4: 5821844-5821844 on Build GRCh38
rs777513481 Chr.4: 5821757-5821757 on Build GRCh38
rs369150626 Chr.4: 5821886-5821886 on Build GRCh38
rs756104100 Chr.4: 5821883-5821883 on Build GRCh38
rs577346929 Chr.4: 5821896-5821896 on Build GRCh38
rs768849755 Chr.4: 5821850-5821850 on Build GRCh38
rs913833703 Chr.4: 5821880-5821880 on Build GRCh38
rs757251245 Chr.4: 5821876-5821876 on Build GRCh38
rs775760953 Chr.4: 5821719-5821719 on Build GRCh38
rs1049251921 Chr.4: 5821802-5821802 on Build GRCh38
rs769164579 Chr.4: 5821902-5821902 on Build GRCh38
rs777646783 Chr.4: 5821832-5821832 on Build GRCh38
rs757302329 Chr.4: 5821834-5821834 on Build GRCh38
rs372825726 Chr.4: 5821868-5821868 on Build GRCh38
rs769178731 Chr.4: 5821870-5821871 on Build GRCh38
rs960998408 Chr.4: 5821908-5821908 on Build GRCh38
rs906240804 Chr.4: 5821907-5821907 on Build GRCh38
rs763173426 Chr.4: 5821805-5821805 on Build GRCh38
rs768959564 Chr.4: 5821889-5821889 on Build GRCh38
rs146955659 Chr.4: 5821843-5821843 on Build GRCh38
rs376553183 Chr.4: 5821858-5821858 on Build GRCh38
rs1035609916 Chr.4: 5821905-5821905 on Build GRCh38
rs769065864 Chr.4: 5821810-5821810 on Build GRCh38
rs987501889 Chr.4: 5821792-5821792 on Build GRCh38
rs747958342 Chr.4: 5821752-5821752 on Build GRCh38
rs746005692 Chr.4: 5821841-5821841 on Build GRCh38
rs762274875 Chr.4: 5821860-5821860 on Build GRCh38
rs765901116 Chr.4: 5821745-5821745 on Build GRCh38
rs763514749 Chr.4: 5821733-5821733 on Build GRCh38
rs910781348 Chr.4: 5821809-5821809 on Build GRCh38
rs762716076 Chr.4: 5821870-5821870 on Build GRCh38
rs781220085 Chr.4: 5821882-5821882 on Build GRCh38
rs770010840 Chr.4: 5821842-5821842 on Build GRCh38
rs572015900 Chr.4: 5821864-5821864 on Build GRCh38
rs750485076 Chr.4: 5821830-5821832 on Build GRCh38
rs372112723 Chr.4: 5821728-5821728 on Build GRCh38
rs765559945 Chr.4: 5821706-5821706 on Build GRCh38
rs763767778 Chr.4: 5821872-5821872 on Build GRCh38
rs758342117 Chr.4: 5821754-5821754 on Build GRCh38
rs765676265 Chr.4: 5821715-5821715 on Build GRCh38
rs754670949 Chr.4: 5821748-5821748 on Build GRCh38
rs981092624 Chr.4: 5821722-5821722 on Build GRCh38
rs779139937 Chr.4: 5821891-5821891 on Build GRCh38
rs764855735 Chr.4: 5821825-5821825 on Build GRCh38
rs535769755 Chr.4: 5821814-5821814 on Build GRCh38
rs759182182 Chr.4: 5821821-5821821 on Build GRCh38
rs755550555 Chr.4: 5821743-5821743 on Build GRCh38
rs751468848 Chr.4: 5821873-5821873 on Build GRCh38
rs764807204 Chr.4: 5821865-5821865 on Build GRCh38
rs765878303 Chr.4: 5821771-5821773 on Build GRCh38
rs201013391 Chr.4: 5821737-5821737 on Build GRCh38
rs780200533 Chr.4: 5821884-5821884 on Build GRCh38
rs1023302639 Chr.4: 5821705-5821705 on Build GRCh38
rs770211777 Chr.4: 5821782-5821782 on Build GRCh38
rs750577945 Chr.4: 5821739-5821739 on Build GRCh38
rs753384463 Chr.4: 5821817-5821817 on Build GRCh38
rs781419836 Chr.4: 5821837-5821837 on Build GRCh38
rs763171037 Chr.4: 5821853-5821854 on Build GRCh38
rs765849718 Chr.4: 5821816-5821816 on Build GRCh38
rs142829070 Chr.4: 5821758-5821758 on Build GRCh38
rs764571830 Chr.4: 5821734-5821734 on Build GRCh38
rs771086650 Chr.4: 5821765-5821765 on Build GRCh38
rs764540850 Chr.4: 5821750-5821750 on Build GRCh38
rs758089833 Chr.4: 5821831-5821831 on Build GRCh38
rs200221510 Chr.4: 5821747-5821747 on Build GRCh38
rs762347676 Chr.4: 5821729-5821729 on Build GRCh38

View Full Product Details