Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154444277-154444525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50A
Entrez Gene ID
9130
Gene Name
family with sequence similarity 50 member A
Gene Aliases
9F, DXS9928E, HXC-26, HXC26, XAP5
UniGene
Hs.54277
Gene Chromosome Location
Chr.X: 154444126-154450654 on Build GRCh38
Transcripts
NM_004699.3

SNP Detail

SNP ID Chromosome Location
rs943892582 Chr.X: 154444448-154444448 on Build GRCh38
rs782292623 Chr.X: 154444388-154444388 on Build GRCh38
rs201400397 Chr.X: 154444389-154444389 on Build GRCh38
rs782310897 Chr.X: 154444458-154444458 on Build GRCh38
rs781968577 Chr.X: 154444394-154444394 on Build GRCh38
rs782307208 Chr.X: 154444393-154444393 on Build GRCh38
rs782229660 Chr.X: 154444449-154444449 on Build GRCh38
rs782428530 Chr.X: 154444353-154444353 on Build GRCh38

View Full Product Details