Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16386052-16386325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15L1
Entrez Gene ID
58513
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases
EPS15R
UniGene
Hs.654639
Gene Chromosome Location
Chr.19: 16355244-16472012 on Build GRCh38
Transcripts
NM_001258374.1 NM_001258375.1 NM_021235.2 NR_047665.1 NR_047666.1 XM_017027086.1 XM_017027087.1 XM_017027088.1 XM_017027089.1 XM_017027090.1 XM_017027091.1 XM_017027092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4783350 c.2106C>T p.L702L Chr.19: 16386229-16386229 on Build GRCh38
5854921 c.2150C>T p.S717F Chr.19: 16386185-16386185 on Build GRCh38
4945869 c.2120C>G p.S707C Chr.19: 16386215-16386215 on Build GRCh38
4617888 c.2130C>T p.P710P Chr.19: 16386205-16386205 on Build GRCh38
4781452 c.2112C>G p.P704P Chr.19: 16386223-16386223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780173380 Chr.19: 16386229-16386229 on Build GRCh38
rs779321753 Chr.19: 16386249-16386249 on Build GRCh38
rs201989047 Chr.19: 16386270-16386270 on Build GRCh38
rs937920102 Chr.19: 16386111-16386111 on Build GRCh38
rs191694456 Chr.19: 16386242-16386242 on Build GRCh38
rs776302300 Chr.19: 16386154-16386154 on Build GRCh38
rs751407853 Chr.19: 16386194-16386194 on Build GRCh38
rs959655019 Chr.19: 16386092-16386092 on Build GRCh38
rs749833602 Chr.19: 16386131-16386131 on Build GRCh38
rs770490994 Chr.19: 16386149-16386149 on Build GRCh38
rs758988763 Chr.19: 16386175-16386175 on Build GRCh38
rs767655570 Chr.19: 16386217-16386217 on Build GRCh38
rs772431004 Chr.19: 16386256-16386256 on Build GRCh38
rs142631693 Chr.19: 16386106-16386106 on Build GRCh38
rs372925875 Chr.19: 16386236-16386236 on Build GRCh38
rs750709081 Chr.19: 16386225-16386225 on Build GRCh38
rs756261464 Chr.19: 16386226-16386226 on Build GRCh38
rs1035006541 Chr.19: 16386085-16386085 on Build GRCh38
rs972601200 Chr.19: 16386228-16386228 on Build GRCh38
rs757038650 Chr.19: 16386199-16386199 on Build GRCh38
rs780087442 Chr.19: 16386129-16386129 on Build GRCh38
rs745604551 Chr.19: 16386271-16386271 on Build GRCh38
rs368429201 Chr.19: 16386145-16386145 on Build GRCh38
rs748486793 Chr.19: 16386148-16386148 on Build GRCh38
rs748488000 Chr.19: 16386251-16386251 on Build GRCh38
rs368728491 Chr.19: 16386110-16386110 on Build GRCh38
rs1054736934 Chr.19: 16386269-16386269 on Build GRCh38
rs763044271 Chr.19: 16386191-16386191 on Build GRCh38
rs775139398 Chr.19: 16386180-16386180 on Build GRCh38
rs1026145606 Chr.19: 16386077-16386077 on Build GRCh38
rs745966861 Chr.19: 16386123-16386123 on Build GRCh38
rs764070106 Chr.19: 16386193-16386193 on Build GRCh38
rs917557262 Chr.19: 16386212-16386212 on Build GRCh38
rs949118670 Chr.19: 16386215-16386215 on Build GRCh38
rs1007780487 Chr.19: 16386276-16386276 on Build GRCh38
rs144918680 Chr.19: 16386138-16386138 on Build GRCh38
rs756171413 Chr.19: 16386127-16386127 on Build GRCh38
rs950548330 Chr.19: 16386081-16386081 on Build GRCh38
rs200435689 Chr.19: 16386274-16386274 on Build GRCh38
rs775157935 Chr.19: 16386278-16386278 on Build GRCh38
rs531717728 Chr.19: 16386118-16386118 on Build GRCh38

View Full Product Details