Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154235666-154235933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS2
Entrez Gene ID
54798
Gene Name
dachsous cadherin-related 2
Gene Aliases
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
UniGene
Hs.655664
Gene Chromosome Location
Chr.4: 154234375-154491778 on Build GRCh38
Transcripts
NM_017639.3 XM_011532045.1 XM_011532046.2 XM_011532049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5671782 c.7571C>T p.S2524F Chr.4: 154235716-154235716 on Build GRCh38
6469351 c.7466T>A p.I2489K Chr.4: 154235821-154235821 on Build GRCh38
447521 c.7577C>G p.S2526C Chr.4: 154235710-154235710 on Build GRCh38
5865915 c.7555G>A p.V2519I Chr.4: 154235732-154235732 on Build GRCh38
229710 c.7432G>A p.G2478R Chr.4: 154235855-154235855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143419229 Chr.4: 154235837-154235837 on Build GRCh38
rs776398167 Chr.4: 154235808-154235808 on Build GRCh38
rs749554957 Chr.4: 154235692-154235692 on Build GRCh38
rs751119232 Chr.4: 154235733-154235733 on Build GRCh38
rs748029791 Chr.4: 154235892-154235892 on Build GRCh38
rs924554739 Chr.4: 154235827-154235827 on Build GRCh38
rs770978953 Chr.4: 154235697-154235697 on Build GRCh38
rs746126538 Chr.4: 154235882-154235882 on Build GRCh38
rs769774264 Chr.4: 154235906-154235906 on Build GRCh38
rs149216376 Chr.4: 154235802-154235802 on Build GRCh38
rs201099367 Chr.4: 154235749-154235749 on Build GRCh38
rs866963796 Chr.4: 154235765-154235765 on Build GRCh38
rs766143078 Chr.4: 154235828-154235828 on Build GRCh38
rs775178193 Chr.4: 154235724-154235724 on Build GRCh38
rs948460639 Chr.4: 154235877-154235877 on Build GRCh38
rs551336329 Chr.4: 154235847-154235847 on Build GRCh38
rs745966444 Chr.4: 154235776-154235776 on Build GRCh38
rs753125225 Chr.4: 154235855-154235855 on Build GRCh38
rs186377292 Chr.4: 154235821-154235821 on Build GRCh38
rs764925333 Chr.4: 154235811-154235811 on Build GRCh38
rs756439016 Chr.4: 154235724-154235725 on Build GRCh38
rs141998004 Chr.4: 154235871-154235871 on Build GRCh38
rs772019908 Chr.4: 154235887-154235887 on Build GRCh38
rs1044202976 Chr.4: 154235886-154235886 on Build GRCh38
rs550433116 Chr.4: 154235688-154235688 on Build GRCh38
rs539466761 Chr.4: 154235751-154235751 on Build GRCh38
rs542037981 Chr.4: 154235748-154235748 on Build GRCh38
rs778252944 Chr.4: 154235769-154235769 on Build GRCh38
rs370459330 Chr.4: 154235890-154235890 on Build GRCh38
rs765982530 Chr.4: 154235727-154235727 on Build GRCh38
rs756705093 Chr.4: 154235856-154235856 on Build GRCh38
rs562183685 Chr.4: 154235721-154235721 on Build GRCh38
rs61743677 Chr.4: 154235863-154235863 on Build GRCh38
rs763948067 Chr.4: 154235720-154235720 on Build GRCh38
rs778000742 Chr.4: 154235732-154235733 on Build GRCh38
rs771214038 Chr.4: 154235775-154235775 on Build GRCh38
rs144401140 Chr.4: 154235685-154235685 on Build GRCh38
rs201587539 Chr.4: 154235735-154235735 on Build GRCh38
rs762627103 Chr.4: 154235822-154235822 on Build GRCh38
rs199850135 Chr.4: 154235870-154235870 on Build GRCh38
rs749501585 Chr.4: 154235774-154235774 on Build GRCh38
rs760604777 Chr.4: 154235705-154235705 on Build GRCh38
rs761858028 Chr.4: 154235810-154235810 on Build GRCh38
rs537787483 Chr.4: 154235866-154235866 on Build GRCh38
rs1057221009 Chr.4: 154235864-154235864 on Build GRCh38
rs199574007 Chr.4: 154235700-154235700 on Build GRCh38
rs751064423 Chr.4: 154235829-154235829 on Build GRCh38
rs61746101 Chr.4: 154235723-154235723 on Build GRCh38
rs1015675487 Chr.4: 154235880-154235880 on Build GRCh38

View Full Product Details