Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 116084942-116085171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD10
Entrez Gene ID
51397
Gene Name
COMM domain containing 10
Gene Aliases
PTD002
UniGene
Hs.483136
Gene Chromosome Location
Chr.5: 116084592-116446462 on Build GRCh38
Transcripts
NM_016144.3 XM_011543458.2 XM_011543460.2 XM_017009555.1 XM_017009556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391089 c.10C>T p.P4S Chr.5: 116085062-116085062 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768516305 Chr.5: 116085048-116085048 on Build GRCh38
rs925746772 Chr.5: 116085050-116085050 on Build GRCh38
rs115091948 Chr.5: 116084963-116084963 on Build GRCh38
rs750827289 Chr.5: 116085128-116085128 on Build GRCh38
rs779596729 Chr.5: 116085040-116085040 on Build GRCh38
rs376346392 Chr.5: 116085081-116085081 on Build GRCh38
rs777007796 Chr.5: 116085015-116085015 on Build GRCh38
rs748135375 Chr.5: 116085141-116085141 on Build GRCh38
rs753532356 Chr.5: 116085137-116085137 on Build GRCh38
rs774293617 Chr.5: 116085120-116085120 on Build GRCh38
rs768789673 Chr.5: 116085116-116085116 on Build GRCh38
rs775881946 Chr.5: 116085100-116085100 on Build GRCh38
rs894944869 Chr.5: 116085059-116085059 on Build GRCh38
rs778689883 Chr.5: 116085115-116085116 on Build GRCh38
rs773187074 Chr.5: 116085090-116085090 on Build GRCh38
rs367851560 Chr.5: 116085014-116085014 on Build GRCh38
rs367887177 Chr.5: 116085026-116085026 on Build GRCh38
rs746765415 Chr.5: 116085092-116085092 on Build GRCh38
rs549127876 Chr.5: 116085151-116085151 on Build GRCh38
rs765538117 Chr.5: 116085065-116085065 on Build GRCh38
rs747381452 Chr.5: 116085006-116085006 on Build GRCh38
rs114781327 Chr.5: 116085016-116085016 on Build GRCh38
rs966511483 Chr.5: 116084995-116084995 on Build GRCh38
rs779650552 Chr.5: 116085080-116085080 on Build GRCh38
rs757640569 Chr.5: 116085074-116085074 on Build GRCh38
rs752814056 Chr.5: 116085023-116085023 on Build GRCh38
rs754581352 Chr.5: 116085138-116085138 on Build GRCh38
rs762442256 Chr.5: 116085124-116085124 on Build GRCh38
rs747978857 Chr.5: 116085055-116085055 on Build GRCh38
rs542755921 Chr.5: 116084977-116084977 on Build GRCh38
rs576054779 Chr.5: 116084996-116084996 on Build GRCh38
rs763032209 Chr.5: 116085113-116085113 on Build GRCh38
rs754842131 Chr.5: 116085096-116085108 on Build GRCh38
rs1004996499 Chr.5: 116084999-116084999 on Build GRCh38
rs1004891565 Chr.5: 116084981-116084981 on Build GRCh38
rs765482969 Chr.5: 116085019-116085019 on Build GRCh38
rs372722098 Chr.5: 116085027-116085027 on Build GRCh38
rs748363200 Chr.5: 116085123-116085123 on Build GRCh38
rs998250659 Chr.5: 116085109-116085109 on Build GRCh38
rs778217936 Chr.5: 116085083-116085083 on Build GRCh38
rs772119143 Chr.5: 116085088-116085088 on Build GRCh38
rs780647238 Chr.5: 116085013-116085013 on Build GRCh38
rs73269257 Chr.5: 116084987-116084987 on Build GRCh38
rs201640787 Chr.5: 116085028-116085028 on Build GRCh38
rs748773522 Chr.5: 116085046-116085046 on Build GRCh38
rs188045129 Chr.5: 116085024-116085024 on Build GRCh38
rs755756992 Chr.5: 116085039-116085039 on Build GRCh38
rs774912902 Chr.5: 116085010-116085010 on Build GRCh38
rs544971587 Chr.5: 116084997-116084997 on Build GRCh38
rs774368450 Chr.5: 116085052-116085052 on Build GRCh38
rs375958593 Chr.5: 116085136-116085136 on Build GRCh38
rs545406867 Chr.5: 116085057-116085057 on Build GRCh38
rs775757089 Chr.5: 116085067-116085067 on Build GRCh38
rs375740112 Chr.5: 116085007-116085007 on Build GRCh38
rs764035865 Chr.5: 116085072-116085072 on Build GRCh38
rs767991573 Chr.5: 116085125-116085125 on Build GRCh38
rs778541311 Chr.5: 116085145-116085145 on Build GRCh38
rs745521984 Chr.5: 116085037-116085037 on Build GRCh38
rs778060416 Chr.5: 116085003-116085003 on Build GRCh38
rs767938566 Chr.5: 116085076-116085076 on Build GRCh38
rs770804923 Chr.5: 116085093-116085093 on Build GRCh38
rs1014913916 Chr.5: 116085148-116085148 on Build GRCh38
rs899464464 Chr.5: 116084978-116084978 on Build GRCh38
rs759898407 Chr.5: 116085062-116085062 on Build GRCh38
rs997865603 Chr.5: 116084983-116084983 on Build GRCh38
rs745471752 Chr.5: 116085008-116085008 on Build GRCh38
rs769304464 Chr.5: 116085009-116085009 on Build GRCh38
rs530307368 Chr.5: 116085085-116085085 on Build GRCh38
rs755814048 Chr.5: 116085077-116085077 on Build GRCh38
rs758972783 Chr.5: 116085002-116085002 on Build GRCh38
rs773139726 Chr.5: 116085058-116085058 on Build GRCh38
rs1005070521 Chr.5: 116085149-116085149 on Build GRCh38
rs752124091 Chr.5: 116085073-116085073 on Build GRCh38
rs999923346 Chr.5: 116084962-116084962 on Build GRCh38

View Full Product Details