Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58645833-58646102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT1
Entrez Gene ID
51339
Gene Name
dishevelled binding antagonist of beta catenin 1
Gene Aliases
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3
UniGene
Hs.48950
Gene Chromosome Location
Chr.14: 58634068-58648321 on Build GRCh38
Transcripts
NM_001079520.1 NM_016651.5 NR_046093.1 NR_046095.1 XM_006720167.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4916900 c.1256G>T p.W419L Chr.14: 58645879-58645879 on Build GRCh38
6140816 c.1293G>A p.K431K Chr.14: 58645916-58645916 on Build GRCh38
4934376 c.1417G>T p.E473* Chr.14: 58646040-58646040 on Build GRCh38
6140815 c.1414G>T p.G472W Chr.14: 58646037-58646037 on Build GRCh38
4902062 c.1444T>G p.S482A Chr.14: 58646067-58646067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749374751 Chr.14: 58646031-58646031 on Build GRCh38
rs753365048 Chr.14: 58645951-58645951 on Build GRCh38
rs781173883 Chr.14: 58645880-58645880 on Build GRCh38
rs761831818 Chr.14: 58645905-58645905 on Build GRCh38
rs772150847 Chr.14: 58646044-58646044 on Build GRCh38
rs375814215 Chr.14: 58646016-58646016 on Build GRCh38
rs34051866 Chr.14: 58646044-58646044 on Build GRCh38
rs1049868053 Chr.14: 58646080-58646080 on Build GRCh38
rs773255963 Chr.14: 58645918-58645918 on Build GRCh38
rs975319593 Chr.14: 58645900-58645900 on Build GRCh38
rs768761714 Chr.14: 58646076-58646081 on Build GRCh38
rs574317881 Chr.14: 58646029-58646029 on Build GRCh38
rs771152948 Chr.14: 58645978-58645978 on Build GRCh38
rs780338769 Chr.14: 58645877-58645882 on Build GRCh38
rs747368131 Chr.14: 58645899-58645899 on Build GRCh38
rs763203257 Chr.14: 58645922-58645922 on Build GRCh38
rs763931609 Chr.14: 58646003-58646003 on Build GRCh38
rs767359253 Chr.14: 58645928-58645928 on Build GRCh38
rs34015825 Chr.14: 58645959-58645959 on Build GRCh38
rs959453627 Chr.14: 58645968-58645968 on Build GRCh38
rs760332340 Chr.14: 58646051-58646051 on Build GRCh38
rs199932678 Chr.14: 58645857-58645857 on Build GRCh38
rs372840266 Chr.14: 58645937-58645937 on Build GRCh38
rs774490030 Chr.14: 58645987-58645987 on Build GRCh38
rs752636968 Chr.14: 58645943-58645943 on Build GRCh38
rs748896918 Chr.14: 58645896-58645896 on Build GRCh38
rs781572728 Chr.14: 58645960-58645960 on Build GRCh38
rs766582645 Chr.14: 58645924-58645924 on Build GRCh38
rs912224905 Chr.14: 58645890-58645890 on Build GRCh38
rs868085760 Chr.14: 58645891-58645892 on Build GRCh38
rs758211732 Chr.14: 58645858-58645858 on Build GRCh38
rs752409499 Chr.14: 58645998-58645998 on Build GRCh38
rs748738357 Chr.14: 58645962-58645962 on Build GRCh38
rs17832998 Chr.14: 58646014-58646014 on Build GRCh38
rs924506374 Chr.14: 58646042-58646042 on Build GRCh38
rs773348564 Chr.14: 58645974-58645974 on Build GRCh38
rs763006263 Chr.14: 58645977-58645977 on Build GRCh38
rs368082534 Chr.14: 58645992-58645992 on Build GRCh38
rs755597010 Chr.14: 58645946-58645946 on Build GRCh38
rs563561287 Chr.14: 58646057-58646057 on Build GRCh38
rs754974285 Chr.14: 58645877-58645877 on Build GRCh38
rs776674599 Chr.14: 58646064-58646064 on Build GRCh38
rs1027144588 Chr.14: 58646036-58646036 on Build GRCh38
rs541714246 Chr.14: 58646041-58646041 on Build GRCh38
rs757031245 Chr.14: 58645953-58645953 on Build GRCh38
rs760704027 Chr.14: 58646002-58646002 on Build GRCh38
rs770956671 Chr.14: 58646033-58646033 on Build GRCh38
rs747248389 Chr.14: 58645895-58645896 on Build GRCh38
rs114859205 Chr.14: 58645927-58645927 on Build GRCh38
rs755583995 Chr.14: 58645883-58645883 on Build GRCh38
rs769869957 Chr.14: 58645908-58645908 on Build GRCh38
rs774575638 Chr.14: 58646039-58646039 on Build GRCh38
rs79977627 Chr.14: 58645902-58645902 on Build GRCh38
rs773769153 Chr.14: 58646018-58646018 on Build GRCh38
rs376695215 Chr.14: 58645925-58645925 on Build GRCh38
rs748458975 Chr.14: 58646025-58646025 on Build GRCh38
rs778652355 Chr.14: 58645958-58645958 on Build GRCh38
rs770682231 Chr.14: 58645898-58645898 on Build GRCh38
rs200763989 Chr.14: 58645950-58645950 on Build GRCh38
rs534806469 Chr.14: 58645990-58645990 on Build GRCh38
rs747625120 Chr.14: 58645882-58645882 on Build GRCh38
rs775419126 Chr.14: 58646046-58646046 on Build GRCh38
rs770172050 Chr.14: 58645966-58645966 on Build GRCh38
rs965230880 Chr.14: 58645892-58645892 on Build GRCh38
rs79146796 Chr.14: 58645981-58645981 on Build GRCh38
rs373688563 Chr.14: 58645870-58645870 on Build GRCh38
rs781652020 Chr.14: 58646024-58646024 on Build GRCh38
rs866279809 Chr.14: 58645942-58645942 on Build GRCh38
rs777133070 Chr.14: 58645903-58645903 on Build GRCh38
rs112117475 Chr.14: 58645926-58645926 on Build GRCh38
rs757965335 Chr.14: 58646023-58646023 on Build GRCh38
rs201114969 Chr.14: 58645871-58645871 on Build GRCh38

View Full Product Details