Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138137060-138137333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135B
Entrez Gene ID
51059
Gene Name
family with sequence similarity 135 member B
Gene Aliases
C8ORFK32
UniGene
Hs.126024
Gene Chromosome Location
Chr.8: 138130023-138498390 on Build GRCh38
Transcripts
NM_015912.3 XM_011517056.2 XM_011517058.2 XM_011517059.1 XM_011517060.2 XM_011517061.2 XM_011517062.1 XM_011517063.2 XM_011517064.2 XM_011517065.1 XM_011517068.2 XM_011517069.2 XM_011517072.2 XM_011517073.2 XM_011517074.1 XM_017013470.1 XM_017013471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
320242 c.4013C>A p.T1338K Chr.8: 138137149-138137149 on Build GRCh38
3645411 c.3903G>A p.G1301G Chr.8: 138137259-138137259 on Build GRCh38
320243 c.4013C>A p.T1338K Chr.8: 138137149-138137149 on Build GRCh38
6988842 c.3902G>A p.G1301E Chr.8: 138137260-138137260 on Build GRCh38
5431855 c.3999C>G p.L1333L Chr.8: 138137163-138137163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376657625 Chr.8: 138137274-138137274 on Build GRCh38
rs745362333 Chr.8: 138137135-138137135 on Build GRCh38
rs940166362 Chr.8: 138137169-138137169 on Build GRCh38
rs6577877 Chr.8: 138137267-138137267 on Build GRCh38
rs746890097 Chr.8: 138137106-138137106 on Build GRCh38
rs374776113 Chr.8: 138137170-138137170 on Build GRCh38
rs777739383 Chr.8: 138137096-138137096 on Build GRCh38
rs746954728 Chr.8: 138137238-138137238 on Build GRCh38
rs757269869 Chr.8: 138137111-138137111 on Build GRCh38
rs748925477 Chr.8: 138137161-138137161 on Build GRCh38
rs772562293 Chr.8: 138137205-138137205 on Build GRCh38
rs568832823 Chr.8: 138137090-138137090 on Build GRCh38
rs1003217373 Chr.8: 138137203-138137203 on Build GRCh38
rs369932293 Chr.8: 138137124-138137124 on Build GRCh38
rs375406846 Chr.8: 138137095-138137095 on Build GRCh38
rs999762436 Chr.8: 138137279-138137279 on Build GRCh38
rs751399732 Chr.8: 138137256-138137256 on Build GRCh38
rs200762412 Chr.8: 138137285-138137285 on Build GRCh38
rs775179492 Chr.8: 138137151-138137151 on Build GRCh38
rs1018872925 Chr.8: 138137280-138137280 on Build GRCh38
rs762240525 Chr.8: 138137189-138137189 on Build GRCh38
rs1008861857 Chr.8: 138137246-138137246 on Build GRCh38
rs748914173 Chr.8: 138137286-138137286 on Build GRCh38
rs932717496 Chr.8: 138137094-138137094 on Build GRCh38
rs6577876 Chr.8: 138137163-138137163 on Build GRCh38
rs552229283 Chr.8: 138137087-138137087 on Build GRCh38
rs369757079 Chr.8: 138137275-138137275 on Build GRCh38
rs542216692 Chr.8: 138137241-138137241 on Build GRCh38
rs576943673 Chr.8: 138137290-138137290 on Build GRCh38
rs778603759 Chr.8: 138137293-138137293 on Build GRCh38
rs902370033 Chr.8: 138137278-138137278 on Build GRCh38
rs867092020 Chr.8: 138137187-138137187 on Build GRCh38
rs759696683 Chr.8: 138137240-138137240 on Build GRCh38
rs150527168 Chr.8: 138137220-138137220 on Build GRCh38
rs769239998 Chr.8: 138137137-138137137 on Build GRCh38
rs773765441 Chr.8: 138137207-138137207 on Build GRCh38
rs748499407 Chr.8: 138137307-138137307 on Build GRCh38
rs766407169 Chr.8: 138137228-138137228 on Build GRCh38

View Full Product Details