Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3508242-3508515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUAP1
Entrez Gene ID
23059
Gene Name
clusterin associated protein 1
Gene Aliases
CFAP22, FAP22, IFT38
UniGene
Hs.155995
Gene Chromosome Location
Chr.16: 3495562-3539048 on Build GRCh38
Transcripts
NM_015041.2 XM_006720867.3 XM_017023068.1 XM_017023069.1 XM_017023070.1 XM_017023071.1 XM_017023072.1 XM_017023073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817947 c.371A>G p.N124S Chr.16: 3508440-3508440 on Build GRCh38
6630841 c.232C>T p.H78Y Chr.16: 3508301-3508301 on Build GRCh38
1377748 c.278C>T p.A93V Chr.16: 3508347-3508347 on Build GRCh38
4486876 c.310C>T p.L104F Chr.16: 3508379-3508379 on Build GRCh38
6630840 c.220-8delA p.? Chr.16: 3508281-3508281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760866222 Chr.16: 3508397-3508397 on Build GRCh38
rs777479822 Chr.16: 3508317-3508317 on Build GRCh38
rs11554091 Chr.16: 3508381-3508381 on Build GRCh38
rs749214234 Chr.16: 3508318-3508318 on Build GRCh38
rs755785058 Chr.16: 3508427-3508427 on Build GRCh38
rs947609181 Chr.16: 3508374-3508374 on Build GRCh38
rs79133712 Chr.16: 3508389-3508389 on Build GRCh38
rs753227494 Chr.16: 3508440-3508440 on Build GRCh38
rs200244691 Chr.16: 3508423-3508423 on Build GRCh38
rs531380218 Chr.16: 3508347-3508347 on Build GRCh38
rs746839973 Chr.16: 3508496-3508496 on Build GRCh38
rs1031747587 Chr.16: 3508387-3508387 on Build GRCh38
rs756795520 Chr.16: 3508458-3508458 on Build GRCh38
rs750707833 Chr.16: 3508360-3508360 on Build GRCh38
rs370512056 Chr.16: 3508342-3508342 on Build GRCh38
rs184112939 Chr.16: 3508422-3508422 on Build GRCh38
rs765807717 Chr.16: 3508357-3508357 on Build GRCh38
rs775322565 Chr.16: 3508395-3508395 on Build GRCh38
rs112533443 Chr.16: 3508460-3508460 on Build GRCh38
rs770939011 Chr.16: 3508323-3508323 on Build GRCh38
rs577364399 Chr.16: 3508380-3508380 on Build GRCh38
rs918143214 Chr.16: 3508403-3508403 on Build GRCh38
rs201336000 Chr.16: 3508394-3508394 on Build GRCh38
rs190930845 Chr.16: 3508293-3508293 on Build GRCh38
rs368448622 Chr.16: 3508447-3508447 on Build GRCh38
rs769420530 Chr.16: 3508314-3508314 on Build GRCh38
rs59492947 Chr.16: 3508283-3508283 on Build GRCh38
rs752380733 Chr.16: 3508377-3508377 on Build GRCh38
rs780345217 Chr.16: 3508376-3508376 on Build GRCh38
rs774132540 Chr.16: 3508465-3508465 on Build GRCh38
rs763859646 Chr.16: 3508435-3508435 on Build GRCh38
rs765608293 Chr.16: 3508421-3508421 on Build GRCh38
rs764147420 Chr.16: 3508349-3508349 on Build GRCh38
rs762197966 Chr.16: 3508354-3508354 on Build GRCh38
rs768663992 Chr.16: 3508407-3508407 on Build GRCh38
rs758318749 Chr.16: 3508489-3508489 on Build GRCh38
rs367665366 Chr.16: 3508492-3508492 on Build GRCh38
rs776654615 Chr.16: 3508406-3508406 on Build GRCh38
rs759395759 Chr.16: 3508340-3508340 on Build GRCh38
rs370605976 Chr.16: 3508398-3508398 on Build GRCh38
rs770723847 Chr.16: 3508383-3508383 on Build GRCh38
rs377209515 Chr.16: 3508378-3508378 on Build GRCh38
rs912929687 Chr.16: 3508365-3508365 on Build GRCh38
rs780924173 Chr.16: 3508304-3508304 on Build GRCh38
rs745581964 Chr.16: 3508391-3508391 on Build GRCh38
rs78524640 Chr.16: 3508393-3508393 on Build GRCh38
rs773976814 Chr.16: 3508338-3508338 on Build GRCh38
rs200792429 Chr.16: 3508425-3508425 on Build GRCh38
rs758808812 Chr.16: 3508366-3508366 on Build GRCh38
rs531848813 Chr.16: 3508449-3508449 on Build GRCh38
rs745652641 Chr.16: 3508441-3508446 on Build GRCh38
rs745494018 Chr.16: 3508332-3508332 on Build GRCh38
rs549707972 Chr.16: 3508490-3508490 on Build GRCh38
rs374595300 Chr.16: 3508488-3508488 on Build GRCh38
rs771973556 Chr.16: 3508479-3508480 on Build GRCh38
rs74751146 Chr.16: 3508274-3508275 on Build GRCh38
rs769586346 Chr.16: 3508348-3508348 on Build GRCh38

View Full Product Details