Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49247344-49247617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC22
Entrez Gene ID
28952
Gene Name
coiled-coil domain containing 22
Gene Aliases
CXorf37, JM1, RTSC2
UniGene
Hs.26333
Gene Chromosome Location
Chr.X: 49235467-49250526 on Build GRCh38
Transcripts
NM_014008.4 XM_005272599.3

SNP Detail

SNP ID Chromosome Location
rs782099905 Chr.X: 49247481-49247481 on Build GRCh38
rs782769431 Chr.X: 49247545-49247545 on Build GRCh38
rs781947521 Chr.X: 49247517-49247517 on Build GRCh38
rs782057751 Chr.X: 49247520-49247520 on Build GRCh38
rs989359760 Chr.X: 49247514-49247514 on Build GRCh38
rs781984730 Chr.X: 49247467-49247467 on Build GRCh38
rs1050641734 Chr.X: 49247388-49247388 on Build GRCh38
rs187851861 Chr.X: 49247578-49247578 on Build GRCh38
rs926510206 Chr.X: 49247482-49247482 on Build GRCh38
rs889365857 Chr.X: 49247389-49247389 on Build GRCh38
rs782173586 Chr.X: 49247574-49247574 on Build GRCh38
rs998080782 Chr.X: 49247379-49247379 on Build GRCh38
rs782576590 Chr.X: 49247565-49247565 on Build GRCh38
rs183877705 Chr.X: 49247512-49247512 on Build GRCh38
rs145051197 Chr.X: 49247408-49247408 on Build GRCh38
rs782516916 Chr.X: 49247544-49247544 on Build GRCh38
rs191242451 Chr.X: 49247402-49247402 on Build GRCh38
rs782818101 Chr.X: 49247528-49247528 on Build GRCh38
rs970526378 Chr.X: 49247444-49247444 on Build GRCh38
rs782632876 Chr.X: 49247443-49247443 on Build GRCh38
rs140735182 Chr.X: 49247548-49247548 on Build GRCh38
rs993214937 Chr.X: 49247413-49247413 on Build GRCh38
rs587780297 Chr.X: 49247558-49247558 on Build GRCh38
rs781882275 Chr.X: 49247542-49247542 on Build GRCh38
rs782152405 Chr.X: 49247403-49247403 on Build GRCh38
rs781830679 Chr.X: 49247547-49247547 on Build GRCh38

View Full Product Details