Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26244115-26244380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP85
Entrez Gene ID
64793
Gene Name
centrosomal protein 85
Gene Aliases
CCDC21
UniGene
Hs.109051
Gene Chromosome Location
Chr.1: 26234153-26278810 on Build GRCh38
Transcripts
NM_001281517.2 NM_001319944.1 NM_022778.4 XM_017002105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6009169 c.128G>A p.R43Q Chr.1: 26244238-26244238 on Build GRCh38
5443974 c.84C>A p.S28S Chr.1: 26244194-26244194 on Build GRCh38
369715 c.88G>T p.G30W Chr.1: 26244198-26244198 on Build GRCh38
3487468 c.83C>T p.S28F Chr.1: 26244193-26244193 on Build GRCh38
7172994 c.160G>A p.D54N Chr.1: 26244270-26244270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753258939 Chr.1: 26244273-26244273 on Build GRCh38
rs756605710 Chr.1: 26244280-26244280 on Build GRCh38
rs986923682 Chr.1: 26244147-26244147 on Build GRCh38
rs560353875 Chr.1: 26244252-26244252 on Build GRCh38
rs771135200 Chr.1: 26244321-26244321 on Build GRCh38
rs963433197 Chr.1: 26244294-26244294 on Build GRCh38
rs200253875 Chr.1: 26244295-26244295 on Build GRCh38
rs757867401 Chr.1: 26244348-26244349 on Build GRCh38
rs780320102 Chr.1: 26244223-26244223 on Build GRCh38
rs540554203 Chr.1: 26244205-26244205 on Build GRCh38
rs768746020 Chr.1: 26244242-26244242 on Build GRCh38
rs776749825 Chr.1: 26244244-26244244 on Build GRCh38
rs769707744 Chr.1: 26244323-26244323 on Build GRCh38
rs556866666 Chr.1: 26244313-26244313 on Build GRCh38
rs1006349497 Chr.1: 26244271-26244271 on Build GRCh38
rs200955763 Chr.1: 26244171-26244171 on Build GRCh38
rs368381610 Chr.1: 26244237-26244237 on Build GRCh38
rs986933431 Chr.1: 26244350-26244350 on Build GRCh38
rs115133994 Chr.1: 26244269-26244269 on Build GRCh38
rs140558906 Chr.1: 26244243-26244243 on Build GRCh38
rs372711332 Chr.1: 26244335-26244335 on Build GRCh38
rs139903267 Chr.1: 26244166-26244166 on Build GRCh38
rs749907422 Chr.1: 26244272-26244272 on Build GRCh38
rs142564361 Chr.1: 26244206-26244206 on Build GRCh38
rs374828034 Chr.1: 26244284-26244284 on Build GRCh38
rs762479134 Chr.1: 26244330-26244330 on Build GRCh38
rs369652785 Chr.1: 26244270-26244270 on Build GRCh38
rs753895345 Chr.1: 26244291-26244291 on Build GRCh38
rs569275098 Chr.1: 26244221-26244221 on Build GRCh38
rs771115123 Chr.1: 26244226-26244226 on Build GRCh38
rs112127457 Chr.1: 26244227-26244227 on Build GRCh38
rs372664199 Chr.1: 26244139-26244139 on Build GRCh38
rs762520687 Chr.1: 26244262-26244262 on Build GRCh38
rs777893077 Chr.1: 26244215-26244215 on Build GRCh38
rs945426766 Chr.1: 26244204-26244204 on Build GRCh38
rs749537587 Chr.1: 26244217-26244217 on Build GRCh38
rs749515523 Chr.1: 26244314-26244314 on Build GRCh38
rs759173668 Chr.1: 26244349-26244349 on Build GRCh38
rs774036383 Chr.1: 26244338-26244338 on Build GRCh38
rs376924614 Chr.1: 26244336-26244336 on Build GRCh38
rs35831900 Chr.1: 26244253-26244253 on Build GRCh38
rs955621109 Chr.1: 26244340-26244340 on Build GRCh38
rs375290306 Chr.1: 26244285-26244285 on Build GRCh38
rs751889433 Chr.1: 26244167-26244167 on Build GRCh38
rs766002558 Chr.1: 26244264-26244264 on Build GRCh38
rs761822338 Chr.1: 26244250-26244250 on Build GRCh38
rs755493447 Chr.1: 26244170-26244170 on Build GRCh38
rs377136346 Chr.1: 26244178-26244178 on Build GRCh38
rs746057005 Chr.1: 26244296-26244296 on Build GRCh38
rs933528544 Chr.1: 26244138-26244138 on Build GRCh38
rs1043094245 Chr.1: 26244173-26244173 on Build GRCh38
rs772615540 Chr.1: 26244327-26244327 on Build GRCh38
rs757443395 Chr.1: 26244293-26244293 on Build GRCh38
rs778255895 Chr.1: 26244157-26244158 on Build GRCh38

View Full Product Details