Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50289494-50289719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPB1
Entrez Gene ID
2553
Gene Name
GA binding protein transcription factor beta subunit 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
UniGene
Hs.654350
Gene Chromosome Location
Chr.15: 50277192-50355429 on Build GRCh38
Transcripts
NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_005254274.3 XM_011521426.2 XM_017022053.1 XM_017022054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2216532 c.776G>A p.G259D Chr.15: 50289626-50289626 on Build GRCh38
7152178 c.832G>A p.D278N Chr.15: 50289570-50289570 on Build GRCh38
6859818 c.734-5_734-4insT p.? Chr.15: 50289672-50289673 on Build GRCh38
7175101 c.868A>G p.T290A Chr.15: 50289534-50289534 on Build GRCh38
5447143 c.752T>A p.V251E Chr.15: 50289650-50289650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375555413 Chr.15: 50289534-50289534 on Build GRCh38
rs77085532 Chr.15: 50289689-50289689 on Build GRCh38
rs749516251 Chr.15: 50289673-50289674 on Build GRCh38
rs751234390 Chr.15: 50289673-50289673 on Build GRCh38
rs773188265 Chr.15: 50289527-50289527 on Build GRCh38
rs765851641 Chr.15: 50289670-50289670 on Build GRCh38
rs765718426 Chr.15: 50289625-50289625 on Build GRCh38
rs756156742 Chr.15: 50289672-50289673 on Build GRCh38
rs769561330 Chr.15: 50289515-50289515 on Build GRCh38
rs968576524 Chr.15: 50289627-50289627 on Build GRCh38
rs746004217 Chr.15: 50289549-50289549 on Build GRCh38
rs374196509 Chr.15: 50289673-50289673 on Build GRCh38
rs762391326 Chr.15: 50289624-50289624 on Build GRCh38
rs776790033 Chr.15: 50289620-50289620 on Build GRCh38
rs757941828 Chr.15: 50289669-50289669 on Build GRCh38
rs754585025 Chr.15: 50289678-50289678 on Build GRCh38
rs74642872 Chr.15: 50289674-50289674 on Build GRCh38
rs368703007 Chr.15: 50289596-50289596 on Build GRCh38
rs17853538 Chr.15: 50289582-50289582 on Build GRCh38
rs750935358 Chr.15: 50289651-50289651 on Build GRCh38
rs772249858 Chr.15: 50289560-50289560 on Build GRCh38
rs781054895 Chr.15: 50289681-50289681 on Build GRCh38
rs775906598 Chr.15: 50289571-50289571 on Build GRCh38
rs1022938524 Chr.15: 50289622-50289622 on Build GRCh38
rs760934659 Chr.15: 50289586-50289586 on Build GRCh38
rs759893078 Chr.15: 50289661-50289661 on Build GRCh38

View Full Product Details