Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44963026-44963299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM179B
Entrez Gene ID
23116
Gene Name
family with sequence similarity 179 member B
Gene Aliases
KIAA0423
UniGene
Hs.371078
Gene Chromosome Location
Chr.14: 44962190-45074431 on Build GRCh38
Transcripts
NM_001308120.1 NM_015091.3 NR_131765.1 XM_011536571.1 XM_017021098.1 XM_017021099.1 XM_017021100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280969 c.785G>A p.R262Q Chr.14: 44963206-44963206 on Build GRCh38
4554965 c.637G>A p.E213K Chr.14: 44963058-44963058 on Build GRCh38
955765 c.811G>T p.E271* Chr.14: 44963232-44963232 on Build GRCh38
4050865 c.857G>A p.R286Q Chr.14: 44963278-44963278 on Build GRCh38
4050863 c.819T>G p.S273S Chr.14: 44963240-44963240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370437319 Chr.14: 44963195-44963195 on Build GRCh38
rs749828747 Chr.14: 44963241-44963241 on Build GRCh38
rs940942709 Chr.14: 44963141-44963141 on Build GRCh38
rs760719137 Chr.14: 44963068-44963068 on Build GRCh38
rs779299119 Chr.14: 44963258-44963258 on Build GRCh38
rs376358743 Chr.14: 44963165-44963165 on Build GRCh38
rs774550576 Chr.14: 44963256-44963256 on Build GRCh38
rs889014419 Chr.14: 44963174-44963174 on Build GRCh38
rs369821527 Chr.14: 44963071-44963071 on Build GRCh38
rs764619463 Chr.14: 44963196-44963196 on Build GRCh38
rs762405947 Chr.14: 44963228-44963230 on Build GRCh38
rs776233602 Chr.14: 44963162-44963162 on Build GRCh38
rs948084781 Chr.14: 44963175-44963175 on Build GRCh38
rs766719339 Chr.14: 44963099-44963099 on Build GRCh38
rs148819135 Chr.14: 44963100-44963100 on Build GRCh38
rs763405402 Chr.14: 44963116-44963116 on Build GRCh38
rs745560761 Chr.14: 44963122-44963122 on Build GRCh38
rs771549070 Chr.14: 44963138-44963138 on Build GRCh38
rs752851971 Chr.14: 44963203-44963203 on Build GRCh38
rs751796645 Chr.14: 44963182-44963182 on Build GRCh38
rs758485147 Chr.14: 44963092-44963092 on Build GRCh38
rs765876852 Chr.14: 44963247-44963247 on Build GRCh38
rs141155288 Chr.14: 44963276-44963276 on Build GRCh38
rs776623041 Chr.14: 44963272-44963272 on Build GRCh38
rs146515749 Chr.14: 44963263-44963263 on Build GRCh38
rs755101967 Chr.14: 44963101-44963101 on Build GRCh38
rs532292559 Chr.14: 44963123-44963123 on Build GRCh38
rs897694237 Chr.14: 44963120-44963120 on Build GRCh38
rs764100355 Chr.14: 44963143-44963143 on Build GRCh38
rs756510795 Chr.14: 44963233-44963233 on Build GRCh38
rs753701888 Chr.14: 44963076-44963076 on Build GRCh38
rs950104072 Chr.14: 44963063-44963063 on Build GRCh38
rs745478112 Chr.14: 44963259-44963259 on Build GRCh38
rs144734587 Chr.14: 44963277-44963277 on Build GRCh38
rs781458414 Chr.14: 44963107-44963107 on Build GRCh38
rs748581640 Chr.14: 44963109-44963109 on Build GRCh38
rs774799569 Chr.14: 44963265-44963265 on Build GRCh38
rs768105823 Chr.14: 44963268-44963268 on Build GRCh38
rs778355251 Chr.14: 44963239-44963239 on Build GRCh38
rs975064276 Chr.14: 44963064-44963064 on Build GRCh38
rs775091625 Chr.14: 44963140-44963140 on Build GRCh38
rs773990159 Chr.14: 44963118-44963118 on Build GRCh38
rs373487359 Chr.14: 44963115-44963115 on Build GRCh38
rs1052704552 Chr.14: 44963248-44963248 on Build GRCh38
rs142524617 Chr.14: 44963246-44963246 on Build GRCh38
rs761877774 Chr.14: 44963274-44963274 on Build GRCh38
rs145441267 Chr.14: 44963081-44963081 on Build GRCh38

View Full Product Details