Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94051264-94051537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX24
Entrez Gene ID
57062
Gene Name
DEAD-box helicase 24
Gene Aliases
-
UniGene
Hs.510328
Gene Chromosome Location
Chr.14: 94050922-94081212 on Build GRCh38
Transcripts
NM_020414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3886581 c.2398C>T p.L800L Chr.14: 94051373-94051373 on Build GRCh38
5982457 c.2378G>A p.R793H Chr.14: 94051393-94051393 on Build GRCh38
1203230 c.2377C>T p.R793C Chr.14: 94051394-94051394 on Build GRCh38
959102 c.2343A>G p.R781R Chr.14: 94051428-94051428 on Build GRCh38
6852974 c.2445G>C p.K815N Chr.14: 94051326-94051326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767766977 Chr.14: 94051508-94051508 on Build GRCh38
rs45505197 Chr.14: 94051498-94051498 on Build GRCh38
rs370682465 Chr.14: 94051288-94051288 on Build GRCh38
rs764165394 Chr.14: 94051495-94051495 on Build GRCh38
rs754865069 Chr.14: 94051394-94051394 on Build GRCh38
rs552250777 Chr.14: 94051320-94051320 on Build GRCh38
rs757372617 Chr.14: 94051436-94051436 on Build GRCh38
rs568654670 Chr.14: 94051448-94051448 on Build GRCh38
rs766939385 Chr.14: 94051348-94051348 on Build GRCh38
rs753898193 Chr.14: 94051454-94051454 on Build GRCh38
rs749037973 Chr.14: 94051390-94051390 on Build GRCh38
rs1041930146 Chr.14: 94051417-94051417 on Build GRCh38
rs765644935 Chr.14: 94051422-94051422 on Build GRCh38
rs752979202 Chr.14: 94051368-94051368 on Build GRCh38
rs779756351 Chr.14: 94051322-94051322 on Build GRCh38
rs750340843 Chr.14: 94051382-94051382 on Build GRCh38
rs778903584 Chr.14: 94051460-94051460 on Build GRCh38
rs780166607 Chr.14: 94051386-94051386 on Build GRCh38
rs746422708 Chr.14: 94051479-94051479 on Build GRCh38
rs755968614 Chr.14: 94051383-94051383 on Build GRCh38
rs747363567 Chr.14: 94051477-94051478 on Build GRCh38
rs200404796 Chr.14: 94051308-94051308 on Build GRCh38
rs754151329 Chr.14: 94051359-94051359 on Build GRCh38
rs747807712 Chr.14: 94051461-94051461 on Build GRCh38
rs770112060 Chr.14: 94051483-94051483 on Build GRCh38
rs909939185 Chr.14: 94051325-94051325 on Build GRCh38
rs925425981 Chr.14: 94051300-94051300 on Build GRCh38
rs368922022 Chr.14: 94051472-94051472 on Build GRCh38
rs536201407 Chr.14: 94051477-94051477 on Build GRCh38
rs745847508 Chr.14: 94051316-94051316 on Build GRCh38
rs777127239 Chr.14: 94051419-94051419 on Build GRCh38
rs777761095 Chr.14: 94051381-94051381 on Build GRCh38
rs761137194 Chr.14: 94051346-94051346 on Build GRCh38
rs375061762 Chr.14: 94051292-94051292 on Build GRCh38
rs147672109 Chr.14: 94051324-94051324 on Build GRCh38
rs774164236 Chr.14: 94051326-94051326 on Build GRCh38
rs768576218 Chr.14: 94051391-94051391 on Build GRCh38
rs780560052 Chr.14: 94051401-94051403 on Build GRCh38
rs139389411 Chr.14: 94051432-94051432 on Build GRCh38
rs779128937 Chr.14: 94051293-94051293 on Build GRCh38
rs146354201 Chr.14: 94051429-94051429 on Build GRCh38
rs770490548 Chr.14: 94051473-94051473 on Build GRCh38
rs758877167 Chr.14: 94051382-94051387 on Build GRCh38
rs771591025 Chr.14: 94051412-94051412 on Build GRCh38
rs751546083 Chr.14: 94051433-94051433 on Build GRCh38
rs185110755 Chr.14: 94051488-94051488 on Build GRCh38
rs757936690 Chr.14: 94051465-94051465 on Build GRCh38
rs758487764 Chr.14: 94051374-94051374 on Build GRCh38
rs199661745 Chr.14: 94051366-94051366 on Build GRCh38
rs772554599 Chr.14: 94051416-94051416 on Build GRCh38
rs759885273 Chr.14: 94051420-94051420 on Build GRCh38
rs765376866 Chr.14: 94051367-94051367 on Build GRCh38
rs754810856 Chr.14: 94051455-94051455 on Build GRCh38
rs142316084 Chr.14: 94051335-94051335 on Build GRCh38
rs538315812 Chr.14: 94051340-94051340 on Build GRCh38
rs781181924 Chr.14: 94051309-94051309 on Build GRCh38
rs918455393 Chr.14: 94051506-94051506 on Build GRCh38
rs141742192 Chr.14: 94051484-94051484 on Build GRCh38
rs377666769 Chr.14: 94051295-94051295 on Build GRCh38
rs1023248211 Chr.14: 94051492-94051492 on Build GRCh38
rs760253546 Chr.14: 94051418-94051418 on Build GRCh38
rs918412200 Chr.14: 94051314-94051314 on Build GRCh38
rs761795189 Chr.14: 94051500-94051500 on Build GRCh38
rs777491733 Chr.14: 94051467-94051467 on Build GRCh38
rs749129149 Chr.14: 94051323-94051323 on Build GRCh38
rs374237706 Chr.14: 94051393-94051393 on Build GRCh38

View Full Product Details