Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75501721-75501984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASKIN2
Entrez Gene ID
57513
Gene Name
CASK interacting protein 2
Gene Aliases
ANKS5B
UniGene
Hs.274408
Gene Chromosome Location
Chr.17: 75500260-75515583 on Build GRCh38
Transcripts
NM_001142643.2 NM_020753.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465165 c.3268G>C p.A1090P Chr.17: 75501806-75501806 on Build GRCh38
7210815 c.3171C>A p.P1057P Chr.17: 75501903-75501903 on Build GRCh38
4390546 c.3115C>G p.P1039A Chr.17: 75501959-75501959 on Build GRCh38
1199638 c.3205C>T p.P1069S Chr.17: 75501869-75501869 on Build GRCh38
7263255 c.3261C>T p.A1087A Chr.17: 75501813-75501813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139160407 Chr.17: 75501851-75501851 on Build GRCh38
rs749612816 Chr.17: 75501900-75501900 on Build GRCh38
rs755876731 Chr.17: 75501786-75501786 on Build GRCh38
rs778203020 Chr.17: 75501960-75501960 on Build GRCh38
rs755152685 Chr.17: 75501936-75501936 on Build GRCh38
rs368588112 Chr.17: 75501918-75501918 on Build GRCh38
rs767839500 Chr.17: 75501939-75501939 on Build GRCh38
rs770539355 Chr.17: 75501818-75501818 on Build GRCh38
rs756246991 Chr.17: 75501957-75501957 on Build GRCh38
rs775251118 Chr.17: 75501752-75501753 on Build GRCh38
rs199651519 Chr.17: 75501817-75501817 on Build GRCh38
rs1030056189 Chr.17: 75501964-75501964 on Build GRCh38
rs549340261 Chr.17: 75501902-75501902 on Build GRCh38
rs370742979 Chr.17: 75501760-75501760 on Build GRCh38
rs760278321 Chr.17: 75501914-75501914 on Build GRCh38
rs142355604 Chr.17: 75501812-75501812 on Build GRCh38
rs374681361 Chr.17: 75501883-75501883 on Build GRCh38
rs143069531 Chr.17: 75501882-75501882 on Build GRCh38
rs529580141 Chr.17: 75501803-75501803 on Build GRCh38
rs369001779 Chr.17: 75501921-75501921 on Build GRCh38
rs369746390 Chr.17: 75501747-75501747 on Build GRCh38
rs772645490 Chr.17: 75501764-75501764 on Build GRCh38
rs964842021 Chr.17: 75501923-75501923 on Build GRCh38
rs748327593 Chr.17: 75501889-75501889 on Build GRCh38
rs760384731 Chr.17: 75501772-75501773 on Build GRCh38
rs989126186 Chr.17: 75501742-75501742 on Build GRCh38
rs755849577 Chr.17: 75501740-75501740 on Build GRCh38
rs778382042 Chr.17: 75501898-75501898 on Build GRCh38
rs375934950 Chr.17: 75501894-75501894 on Build GRCh38
rs766089142 Chr.17: 75501750-75501750 on Build GRCh38
rs763909353 Chr.17: 75501915-75501915 on Build GRCh38
rs964456862 Chr.17: 75501746-75501746 on Build GRCh38
rs770077901 Chr.17: 75501739-75501739 on Build GRCh38
rs200708333 Chr.17: 75501887-75501887 on Build GRCh38
rs376483918 Chr.17: 75501816-75501816 on Build GRCh38
rs373169799 Chr.17: 75501748-75501748 on Build GRCh38
rs74385060 Chr.17: 75501785-75501785 on Build GRCh38
rs758677754 Chr.17: 75501797-75501797 on Build GRCh38
rs751765830 Chr.17: 75501935-75501935 on Build GRCh38
rs546501855 Chr.17: 75501863-75501863 on Build GRCh38
rs763724142 Chr.17: 75501795-75501797 on Build GRCh38
rs752685803 Chr.17: 75501956-75501956 on Build GRCh38
rs370336352 Chr.17: 75501860-75501860 on Build GRCh38
rs781689037 Chr.17: 75501813-75501813 on Build GRCh38
rs770131521 Chr.17: 75501815-75501815 on Build GRCh38
rs766946055 Chr.17: 75501841-75501841 on Build GRCh38
rs540478066 Chr.17: 75501762-75501762 on Build GRCh38
rs545113178 Chr.17: 75501842-75501842 on Build GRCh38
rs764148365 Chr.17: 75501859-75501859 on Build GRCh38
rs758998002 Chr.17: 75501840-75501840 on Build GRCh38
rs199506124 Chr.17: 75501905-75501905 on Build GRCh38
rs975846542 Chr.17: 75501927-75501927 on Build GRCh38
rs745520013 Chr.17: 75501903-75501903 on Build GRCh38
rs559845620 Chr.17: 75501862-75501862 on Build GRCh38
rs199907273 Chr.17: 75501814-75501814 on Build GRCh38
rs751044525 Chr.17: 75501752-75501752 on Build GRCh38
rs780475527 Chr.17: 75501802-75501802 on Build GRCh38
rs201735850 Chr.17: 75501751-75501751 on Build GRCh38
rs774846710 Chr.17: 75501910-75501910 on Build GRCh38
rs146308250 Chr.17: 75501825-75501825 on Build GRCh38
rs777430977 Chr.17: 75501795-75501795 on Build GRCh38
rs201276652 Chr.17: 75501848-75501848 on Build GRCh38

View Full Product Details