Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45034054-45034319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAKD
Entrez Gene ID
79877
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
UniGene
Hs.463148
Gene Chromosome Location
Chr.17: 45023338-45061129 on Build GRCh38
Transcripts
NM_001128631.2 NM_001288654.2 NM_001288655.1 NM_001321326.1 NM_024819.6 XM_005257688.2 XM_011525262.2 XM_017025102.1 XM_017025103.1 XM_017025104.1 XM_017025105.1 XM_017025106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1610347 c.252C>T p.N84N Chr.17: 45034251-45034251 on Build GRCh38
5972225 c.213C>T p.D71D Chr.17: 45034290-45034290 on Build GRCh38
1383761 c.196C>T p.R66C Chr.17: 45034307-45034307 on Build GRCh38
7363761 c.239G>A p.R80Q Chr.17: 45034264-45034264 on Build GRCh38
1383760 c.230C>A p.P77H Chr.17: 45034273-45034273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377407699 Chr.17: 45034166-45034166 on Build GRCh38
rs142327107 Chr.17: 45034206-45034206 on Build GRCh38
rs753946708 Chr.17: 45034154-45034154 on Build GRCh38
rs748998128 Chr.17: 45034292-45034292 on Build GRCh38
rs746572447 Chr.17: 45034235-45034235 on Build GRCh38
rs372084014 Chr.17: 45034243-45034243 on Build GRCh38
rs746973107 Chr.17: 45034187-45034187 on Build GRCh38
rs181186960 Chr.17: 45034296-45034296 on Build GRCh38
rs766425411 Chr.17: 45034153-45034153 on Build GRCh38
rs767917991 Chr.17: 45034223-45034223 on Build GRCh38
rs138739628 Chr.17: 45034250-45034250 on Build GRCh38
rs770890283 Chr.17: 45034189-45034189 on Build GRCh38
rs750932586 Chr.17: 45034090-45034090 on Build GRCh38
rs929980690 Chr.17: 45034202-45034202 on Build GRCh38
rs758295915 Chr.17: 45034173-45034173 on Build GRCh38
rs769251277 Chr.17: 45034267-45034268 on Build GRCh38
rs117298907 Chr.17: 45034190-45034190 on Build GRCh38
rs747838296 Chr.17: 45034231-45034232 on Build GRCh38
rs200330941 Chr.17: 45034265-45034265 on Build GRCh38
rs745756989 Chr.17: 45034278-45034278 on Build GRCh38
rs774958081 Chr.17: 45034290-45034290 on Build GRCh38
rs942158905 Chr.17: 45034172-45034172 on Build GRCh38
rs776928993 Chr.17: 45034264-45034264 on Build GRCh38
rs756621108 Chr.17: 45034150-45034150 on Build GRCh38
rs59549223 Chr.17: 45034236-45034236 on Build GRCh38
rs892182530 Chr.17: 45034284-45034284 on Build GRCh38
rs755038775 Chr.17: 45034159-45034159 on Build GRCh38
rs561238574 Chr.17: 45034255-45034255 on Build GRCh38
rs777489214 Chr.17: 45034267-45034267 on Build GRCh38
rs377356588 Chr.17: 45034186-45034186 on Build GRCh38
rs780649662 Chr.17: 45034213-45034214 on Build GRCh38
rs767821589 Chr.17: 45034085-45034085 on Build GRCh38
rs769325051 Chr.17: 45034286-45034286 on Build GRCh38
rs746818832 Chr.17: 45034274-45034274 on Build GRCh38
rs200803823 Chr.17: 45034251-45034251 on Build GRCh38
rs1032849522 Chr.17: 45034269-45034269 on Build GRCh38
rs762553298 Chr.17: 45034215-45034215 on Build GRCh38
rs775146251 Chr.17: 45034211-45034211 on Build GRCh38
rs17850104 Chr.17: 45034252-45034252 on Build GRCh38
rs955782024 Chr.17: 45034097-45034097 on Build GRCh38
rs779287996 Chr.17: 45034165-45034165 on Build GRCh38
rs1054759071 Chr.17: 45034228-45034228 on Build GRCh38
rs769238846 Chr.17: 45034208-45034208 on Build GRCh38
rs923524297 Chr.17: 45034104-45034104 on Build GRCh38
rs148949186 Chr.17: 45034275-45034275 on Build GRCh38
rs777684072 Chr.17: 45034182-45034182 on Build GRCh38

View Full Product Details