Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6502688-6502945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
UniGene
Hs.720080
Gene Chromosome Location
Chr.11: 6497296-6572025 on Build GRCh38
Transcripts
NM_144666.2 NM_173589.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
930404 c.797G>T p.S266I Chr.11: 6502803-6502803 on Build GRCh38
6134032 c.910C>T p.R304W Chr.11: 6502916-6502916 on Build GRCh38
6410342 c.902C>T p.A301V Chr.11: 6502908-6502908 on Build GRCh38
4968828 c.871A>G p.K291E Chr.11: 6502877-6502877 on Build GRCh38
6134033 c.751C>G p.Q251E Chr.11: 6502757-6502757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539207151 Chr.11: 6502916-6502916 on Build GRCh38
rs768884539 Chr.11: 6502836-6502836 on Build GRCh38
rs369158056 Chr.11: 6502800-6502800 on Build GRCh38
rs78466420 Chr.11: 6502896-6502896 on Build GRCh38
rs928725864 Chr.11: 6502889-6502889 on Build GRCh38
rs139180173 Chr.11: 6502759-6502759 on Build GRCh38
rs745347265 Chr.11: 6502799-6502799 on Build GRCh38
rs780778084 Chr.11: 6502749-6502749 on Build GRCh38
rs748956904 Chr.11: 6502903-6502903 on Build GRCh38
rs867909625 Chr.11: 6502792-6502793 on Build GRCh38
rs745828515 Chr.11: 6502907-6502907 on Build GRCh38
rs779556726 Chr.11: 6502899-6502901 on Build GRCh38
rs150283566 Chr.11: 6502904-6502904 on Build GRCh38
rs759402797 Chr.11: 6502786-6502786 on Build GRCh38
rs373358545 Chr.11: 6502849-6502849 on Build GRCh38
rs762146719 Chr.11: 6502810-6502810 on Build GRCh38
rs774329164 Chr.11: 6502781-6502781 on Build GRCh38
rs778622133 Chr.11: 6502905-6502905 on Build GRCh38
rs758913219 Chr.11: 6502872-6502872 on Build GRCh38
rs771959929 Chr.11: 6502909-6502909 on Build GRCh38
rs764265627 Chr.11: 6502807-6502807 on Build GRCh38
rs779615668 Chr.11: 6502717-6502717 on Build GRCh38
rs747846387 Chr.11: 6502750-6502750 on Build GRCh38
rs780869655 Chr.11: 6502882-6502882 on Build GRCh38
rs770690687 Chr.11: 6502779-6502779 on Build GRCh38
rs561124343 Chr.11: 6502808-6502808 on Build GRCh38
rs754606379 Chr.11: 6502738-6502738 on Build GRCh38
rs369544858 Chr.11: 6502917-6502917 on Build GRCh38
rs749076323 Chr.11: 6502767-6502767 on Build GRCh38
rs1041794458 Chr.11: 6502854-6502854 on Build GRCh38
rs746368615 Chr.11: 6502734-6502734 on Build GRCh38
rs769403084 Chr.11: 6502845-6502845 on Build GRCh38
rs11605196 Chr.11: 6502842-6502842 on Build GRCh38
rs149962461 Chr.11: 6502790-6502790 on Build GRCh38
rs769278853 Chr.11: 6502755-6502755 on Build GRCh38
rs775500522 Chr.11: 6502795-6502795 on Build GRCh38
rs755618176 Chr.11: 6502902-6502902 on Build GRCh38

View Full Product Details