Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15017001-15017270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB2
Entrez Gene ID
132864
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
CPE-BP2, CPEB-2, hCPEB-2
UniGene
Hs.656937
Gene Chromosome Location
Chr.4: 15002451-15070153 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1 XM_017007733.1 XM_017007734.1 NR_125911.1

Gene Symbol
LOC101929095
Entrez Gene ID
101929095
Gene Name
uncharacterized LOC101929095
Gene Aliases
-
UniGene
Hs.656908
Gene Chromosome Location
Chr.4: 15004942-15427914 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1 XM_017007733.1 XM_017007734.1 NR_125911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634532 c.799C>T p.H267Y Chr.4: 15017263-15017263 on Build GRCh38
6634531 c.2110C>T p.H704Y Chr.4: 15017263-15017263 on Build GRCh38
3127349 c.742G>T p.D248Y Chr.4: 15017206-15017206 on Build GRCh38
4122663 c.724-1G>A p.? Chr.4: 15017187-15017187 on Build GRCh38
7087318 c.2092G>T p.D698Y Chr.4: 15017245-15017245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564916263 Chr.4: 15017152-15017152 on Build GRCh38
rs113112809 Chr.4: 15017174-15017174 on Build GRCh38
rs574565373 Chr.4: 15017024-15017024 on Build GRCh38
rs545036583 Chr.4: 15017146-15017146 on Build GRCh38
rs541114402 Chr.4: 15017165-15017165 on Build GRCh38
rs748379716 Chr.4: 15017145-15017145 on Build GRCh38
rs954628980 Chr.4: 15017029-15017029 on Build GRCh38
rs527824372 Chr.4: 15017153-15017153 on Build GRCh38
rs543355824 Chr.4: 15017133-15017133 on Build GRCh38
rs777155910 Chr.4: 15017164-15017164 on Build GRCh38
rs557282003 Chr.4: 15017238-15017238 on Build GRCh38
rs1009245569 Chr.4: 15017050-15017050 on Build GRCh38
rs765568603 Chr.4: 15017166-15017166 on Build GRCh38
rs752503858 Chr.4: 15017183-15017184 on Build GRCh38
rs561329953 Chr.4: 15017179-15017179 on Build GRCh38
rs754810826 Chr.4: 15017224-15017224 on Build GRCh38
rs570297550 Chr.4: 15017244-15017244 on Build GRCh38
rs767199319 Chr.4: 15017183-15017183 on Build GRCh38
rs778644757 Chr.4: 15017226-15017226 on Build GRCh38
rs771379729 Chr.4: 15017162-15017162 on Build GRCh38
rs749841188 Chr.4: 15017184-15017184 on Build GRCh38
rs972041365 Chr.4: 15017025-15017025 on Build GRCh38
rs767334000 Chr.4: 15017162-15017163 on Build GRCh38
rs925698336 Chr.4: 15017118-15017118 on Build GRCh38
rs144630360 Chr.4: 15017239-15017239 on Build GRCh38
rs774039745 Chr.4: 15017177-15017177 on Build GRCh38
rs1043423060 Chr.4: 15017208-15017208 on Build GRCh38
rs115893002 Chr.4: 15017135-15017135 on Build GRCh38
rs537202465 Chr.4: 15017033-15017033 on Build GRCh38
rs767462172 Chr.4: 15017102-15017102 on Build GRCh38
rs967599302 Chr.4: 15017070-15017070 on Build GRCh38
rs530269898 Chr.4: 15017198-15017198 on Build GRCh38
rs753878056 Chr.4: 15017202-15017202 on Build GRCh38
rs951996699 Chr.4: 15017052-15017052 on Build GRCh38
rs778804198 Chr.4: 15017141-15017142 on Build GRCh38
rs748608285 Chr.4: 15017159-15017161 on Build GRCh38
rs142029464 Chr.4: 15017134-15017134 on Build GRCh38
rs373869802 Chr.4: 15017163-15017163 on Build GRCh38
rs761519784 Chr.4: 15017100-15017100 on Build GRCh38

View Full Product Details