Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17759775-17760036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEN1
Entrez Gene ID
348654
Gene Name
GEN1, Holliday junction 5' flap endonuclease
Gene Aliases
Gen
UniGene
Hs.467793
Gene Chromosome Location
Chr.2: 17753687-17785920 on Build GRCh38
Transcripts
NM_001130009.2 NM_182625.4 XM_005262613.4 XM_006712005.3 XM_011532820.2 XM_011532821.2 XM_011532822.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3961632 c.47A>G p.H16R Chr.2: 17759990-17759990 on Build GRCh38
4087450 c.76A>C p.T26P Chr.2: 17760019-17760019 on Build GRCh38
1010584 c.89A>G p.D30G Chr.2: 17760032-17760032 on Build GRCh38
6670467 c.73delA p.T26fs*6 Chr.2: 17760016-17760016 on Build GRCh38
1208105 c.27T>G p.I9M Chr.2: 17759970-17759970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007459953 Chr.2: 17759825-17759825 on Build GRCh38
rs913355820 Chr.2: 17759832-17759832 on Build GRCh38
rs748705786 Chr.2: 17759952-17759952 on Build GRCh38
rs981827265 Chr.2: 17759888-17759891 on Build GRCh38
rs777329788 Chr.2: 17759986-17759986 on Build GRCh38
rs199871757 Chr.2: 17759914-17759914 on Build GRCh38
rs550495634 Chr.2: 17759808-17759808 on Build GRCh38
rs759263092 Chr.2: 17759893-17759893 on Build GRCh38
rs1017452265 Chr.2: 17759849-17759849 on Build GRCh38
rs878940800 Chr.2: 17759840-17759840 on Build GRCh38
rs375511356 Chr.2: 17760002-17760002 on Build GRCh38
rs375762702 Chr.2: 17759913-17759913 on Build GRCh38
rs200070178 Chr.2: 17759932-17759932 on Build GRCh38
rs755591349 Chr.2: 17759965-17759965 on Build GRCh38
rs761930824 Chr.2: 17759902-17759902 on Build GRCh38
rs541211554 Chr.2: 17759853-17759853 on Build GRCh38
rs781689053 Chr.2: 17759944-17759944 on Build GRCh38
rs775317310 Chr.2: 17759901-17759901 on Build GRCh38
rs527906661 Chr.2: 17759821-17759821 on Build GRCh38
rs767421696 Chr.2: 17759894-17759894 on Build GRCh38
rs751889298 Chr.2: 17759922-17759922 on Build GRCh38
rs750638789 Chr.2: 17759912-17759912 on Build GRCh38
rs139372712 Chr.2: 17759988-17759988 on Build GRCh38
rs778434447 Chr.2: 17760001-17760001 on Build GRCh38
rs765453698 Chr.2: 17759910-17759910 on Build GRCh38
rs552152015 Chr.2: 17759997-17759997 on Build GRCh38

View Full Product Details