Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96646292-96646559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
UniGene
Hs.534025
Gene Chromosome Location
Chr.2: 96642719-96704887 on Build GRCh38
Transcripts
NM_001293083.1 XM_011512110.2 XM_011512111.2 XM_011512113.2 XM_011512114.2 XM_011512115.2 XM_011512116.2 XM_011512117.2 XM_011512118.2 XM_011512119.2 XM_011512120.2 XM_011512122.2 XM_011512125.2 XM_011512126.2 XM_011512127.2 XM_011512128.2 XM_017005222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913284 c.130T>A p.W44R Chr.2: 96646445-96646445 on Build GRCh38
1023935 c.93G>T p.K31N Chr.2: 96646408-96646408 on Build GRCh38
6046735 c.127G>A p.V43M Chr.2: 96646442-96646442 on Build GRCh38
4913285 c.12T>A p.C4* Chr.2: 96646445-96646445 on Build GRCh38
6046736 c.9G>A p.P3P Chr.2: 96646442-96646442 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969798346 Chr.2: 96646340-96646340 on Build GRCh38
rs186957094 Chr.2: 96646318-96646318 on Build GRCh38
rs1039127722 Chr.2: 96646471-96646471 on Build GRCh38
rs776697779 Chr.2: 96646486-96646488 on Build GRCh38
rs943464174 Chr.2: 96646450-96646450 on Build GRCh38
rs933893005 Chr.2: 96646418-96646418 on Build GRCh38
rs765614142 Chr.2: 96646441-96646441 on Build GRCh38
rs892062225 Chr.2: 96646524-96646524 on Build GRCh38
rs918526287 Chr.2: 96646396-96646396 on Build GRCh38
rs764234083 Chr.2: 96646464-96646464 on Build GRCh38
rs761924602 Chr.2: 96646528-96646528 on Build GRCh38
rs904613468 Chr.2: 96646491-96646491 on Build GRCh38
rs368841036 Chr.2: 96646442-96646442 on Build GRCh38
rs767171458 Chr.2: 96646389-96646389 on Build GRCh38
rs1056639356 Chr.2: 96646446-96646446 on Build GRCh38
rs930377974 Chr.2: 96646338-96646338 on Build GRCh38
rs757483840 Chr.2: 96646499-96646499 on Build GRCh38
rs754135057 Chr.2: 96646410-96646410 on Build GRCh38
rs1014351414 Chr.2: 96646321-96646321 on Build GRCh38
rs374710689 Chr.2: 96646424-96646424 on Build GRCh38
rs373982838 Chr.2: 96646497-96646497 on Build GRCh38
rs920290917 Chr.2: 96646322-96646322 on Build GRCh38
rs747041075 Chr.2: 96646352-96646352 on Build GRCh38
rs972575882 Chr.2: 96646381-96646381 on Build GRCh38
rs1014469642 Chr.2: 96646538-96646538 on Build GRCh38
rs71427088 Chr.2: 96646413-96646413 on Build GRCh38
rs1009833682 Chr.2: 96646503-96646503 on Build GRCh38
rs936195486 Chr.2: 96646494-96646494 on Build GRCh38
rs1016688080 Chr.2: 96646353-96646353 on Build GRCh38
rs1023950786 Chr.2: 96646329-96646329 on Build GRCh38
rs962577522 Chr.2: 96646359-96646359 on Build GRCh38
rs773795581 Chr.2: 96646516-96646516 on Build GRCh38

View Full Product Details