Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35641273-35641534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD8
Entrez Gene ID
8325
Gene Name
frizzled class receptor 8
Gene Aliases
FZ-8, hFZ8
UniGene
Hs.302634
Gene Chromosome Location
Chr.10: 35638249-35641434 on Build GRCh38
Transcripts
NM_031866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685992 c.115A>G p.T39A Chr.10: 35641315-35641315 on Build GRCh38
6668224 c.121C>T p.P41S Chr.10: 35641309-35641309 on Build GRCh38
6239324 c.26T>A p.V9E Chr.10: 35641404-35641404 on Build GRCh38
378619 c.72C>T p.G24G Chr.10: 35641358-35641358 on Build GRCh38
917910 c.89C>T p.A30V Chr.10: 35641341-35641341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569524518 Chr.10: 35641495-35641495 on Build GRCh38
rs781444551 Chr.10: 35641459-35641459 on Build GRCh38
rs1029202117 Chr.10: 35641389-35641389 on Build GRCh38
rs988871537 Chr.10: 35641295-35641295 on Build GRCh38
rs572546710 Chr.10: 35641493-35641494 on Build GRCh38
rs767000197 Chr.10: 35641410-35641410 on Build GRCh38
rs867537655 Chr.10: 35641395-35641395 on Build GRCh38
rs756969164 Chr.10: 35641437-35641437 on Build GRCh38
rs752869092 Chr.10: 35641455-35641455 on Build GRCh38
rs866654695 Chr.10: 35641378-35641378 on Build GRCh38
rs902054406 Chr.10: 35641346-35641346 on Build GRCh38
rs749960442 Chr.10: 35641442-35641442 on Build GRCh38
rs142623421 Chr.10: 35641484-35641485 on Build GRCh38
rs1038491757 Chr.10: 35641487-35641488 on Build GRCh38
rs746788907 Chr.10: 35641447-35641447 on Build GRCh38
rs746809017 Chr.10: 35641385-35641385 on Build GRCh38
rs773101447 Chr.10: 35641471-35641471 on Build GRCh38
rs980652034 Chr.10: 35641510-35641510 on Build GRCh38
rs759953400 Chr.10: 35641478-35641478 on Build GRCh38
rs71003903 Chr.10: 35641486-35641487 on Build GRCh38
rs896453594 Chr.10: 35641490-35641491 on Build GRCh38
rs202043751 Chr.10: 35641450-35641450 on Build GRCh38
rs767737240 Chr.10: 35641479-35641479 on Build GRCh38
rs773372232 Chr.10: 35641403-35641403 on Build GRCh38
rs145725294 Chr.10: 35641398-35641398 on Build GRCh38
rs370207188 Chr.10: 35641439-35641439 on Build GRCh38
rs750941001 Chr.10: 35641316-35641316 on Build GRCh38
rs763377004 Chr.10: 35641319-35641319 on Build GRCh38
rs745755559 Chr.10: 35641362-35641362 on Build GRCh38
rs776771781 Chr.10: 35641396-35641396 on Build GRCh38
rs748277131 Chr.10: 35641464-35641464 on Build GRCh38
rs751988158 Chr.10: 35641324-35641324 on Build GRCh38
rs763503868 Chr.10: 35641405-35641405 on Build GRCh38
rs756770162 Chr.10: 35641350-35641350 on Build GRCh38
rs869102112 Chr.10: 35641485-35641486 on Build GRCh38
rs1014135361 Chr.10: 35641494-35641495 on Build GRCh38
rs763780475 Chr.10: 35641431-35641431 on Build GRCh38
rs770074661 Chr.10: 35641469-35641469 on Build GRCh38
rs1035443085 Chr.10: 35641489-35641490 on Build GRCh38
rs956984979 Chr.10: 35641421-35641421 on Build GRCh38
rs780028961 Chr.10: 35641446-35641446 on Build GRCh38
rs373590168 Chr.10: 35641474-35641474 on Build GRCh38
rs779763289 Chr.10: 35641373-35641373 on Build GRCh38
rs751969710 Chr.10: 35641414-35641414 on Build GRCh38
rs552560418 Chr.10: 35641492-35641493 on Build GRCh38
rs768858685 Chr.10: 35641393-35641393 on Build GRCh38
rs777637119 Chr.10: 35641342-35641342 on Build GRCh38
rs200372688 Chr.10: 35641297-35641297 on Build GRCh38
rs755762525 Chr.10: 35641341-35641341 on Build GRCh38
rs141014511 Chr.10: 35641365-35641365 on Build GRCh38
rs769803369 Chr.10: 35641401-35641401 on Build GRCh38
rs866267454 Chr.10: 35641298-35641298 on Build GRCh38
rs766807354 Chr.10: 35641320-35641320 on Build GRCh38
rs1022830482 Chr.10: 35641507-35641507 on Build GRCh38
rs889965790 Chr.10: 35641451-35641451 on Build GRCh38
rs776703169 Chr.10: 35641304-35641304 on Build GRCh38
rs149765415 Chr.10: 35641367-35641367 on Build GRCh38
rs749447789 Chr.10: 35641472-35641472 on Build GRCh38
rs753476849 Chr.10: 35641432-35641432 on Build GRCh38
rs533033242 Chr.10: 35641314-35641314 on Build GRCh38
rs934250082 Chr.10: 35641415-35641415 on Build GRCh38
rs753375253 Chr.10: 35641344-35641344 on Build GRCh38
rs941500936 Chr.10: 35641475-35641475 on Build GRCh38
rs761860878 Chr.10: 35641307-35641307 on Build GRCh38
rs375976496 Chr.10: 35641370-35641370 on Build GRCh38
rs367641187 Chr.10: 35641477-35641477 on Build GRCh38
rs778882521 Chr.10: 35641359-35641359 on Build GRCh38
rs760037342 Chr.10: 35641423-35641423 on Build GRCh38

View Full Product Details