Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49663008-49663281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBPL
Entrez Gene ID
84650
Gene Name
emopamil binding protein like
Gene Aliases
EBRP
UniGene
Hs.433278
Gene Chromosome Location
Chr.13: 49660676-49691487 on Build GRCh38
Transcripts
NM_001278636.1 NM_032565.4 NR_103802.1 NR_103803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947902 c.261T>G p.A87A Chr.13: 49663176-49663176 on Build GRCh38
1706777 c.273G>A p.W91* Chr.13: 49663164-49663164 on Build GRCh38
3736090 c.257T>A p.V86D Chr.13: 49663162-49663162 on Build GRCh38
6448759 c.348C>T p.L116L Chr.13: 49663089-49663089 on Build GRCh38
6448760 c.330C>T p.L110L Chr.13: 49663089-49663089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374925327 Chr.13: 49663043-49663043 on Build GRCh38
rs760028150 Chr.13: 49663065-49663065 on Build GRCh38
rs779911608 Chr.13: 49663040-49663042 on Build GRCh38
rs780713413 Chr.13: 49663216-49663216 on Build GRCh38
rs763414494 Chr.13: 49663184-49663184 on Build GRCh38
rs541779688 Chr.13: 49663139-49663139 on Build GRCh38
rs368785849 Chr.13: 49663034-49663034 on Build GRCh38
rs1045564570 Chr.13: 49663207-49663207 on Build GRCh38
rs754485255 Chr.13: 49663134-49663139 on Build GRCh38
rs775861935 Chr.13: 49663182-49663182 on Build GRCh38
rs781178537 Chr.13: 49663161-49663161 on Build GRCh38
rs763760311 Chr.13: 49663094-49663094 on Build GRCh38
rs770144938 Chr.13: 49663078-49663078 on Build GRCh38
rs768880129 Chr.13: 49663218-49663218 on Build GRCh38
rs555120414 Chr.13: 49663165-49663165 on Build GRCh38
rs775772027 Chr.13: 49663080-49663080 on Build GRCh38
rs367944926 Chr.13: 49663173-49663173 on Build GRCh38
rs576964816 Chr.13: 49663244-49663244 on Build GRCh38
rs193298283 Chr.13: 49663151-49663151 on Build GRCh38
rs749896547 Chr.13: 49663230-49663230 on Build GRCh38
rs774111178 Chr.13: 49663222-49663222 on Build GRCh38
rs770234835 Chr.13: 49663177-49663177 on Build GRCh38
rs747181524 Chr.13: 49663146-49663146 on Build GRCh38
rs756852952 Chr.13: 49663142-49663142 on Build GRCh38
rs751139616 Chr.13: 49663105-49663105 on Build GRCh38
rs540970487 Chr.13: 49663250-49663250 on Build GRCh38
rs113216911 Chr.13: 49663057-49663057 on Build GRCh38
rs565115273 Chr.13: 49663229-49663229 on Build GRCh38
rs905153562 Chr.13: 49663212-49663212 on Build GRCh38
rs893014688 Chr.13: 49663171-49663171 on Build GRCh38
rs367558500 Chr.13: 49663118-49663118 on Build GRCh38
rs202068114 Chr.13: 49663227-49663227 on Build GRCh38
rs751490657 Chr.13: 49663067-49663070 on Build GRCh38
rs34505073 Chr.13: 49663119-49663119 on Build GRCh38
rs572356171 Chr.13: 49663121-49663121 on Build GRCh38
rs764596735 Chr.13: 49663249-49663249 on Build GRCh38
rs762533287 Chr.13: 49663089-49663089 on Build GRCh38
rs772281787 Chr.13: 49663036-49663036 on Build GRCh38
rs780028849 Chr.13: 49663055-49663055 on Build GRCh38
rs539014275 Chr.13: 49663045-49663045 on Build GRCh38
rs763844736 Chr.13: 49663058-49663058 on Build GRCh38
rs970046231 Chr.13: 49663232-49663232 on Build GRCh38
rs146925740 Chr.13: 49663122-49663122 on Build GRCh38
rs371466213 Chr.13: 49663056-49663056 on Build GRCh38
rs933651747 Chr.13: 49663074-49663074 on Build GRCh38
rs372070210 Chr.13: 49663191-49663191 on Build GRCh38
rs753641175 Chr.13: 49663125-49663125 on Build GRCh38

View Full Product Details