Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202965302-202965575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R1
Entrez Gene ID
51706
Gene Name
cytochrome b5 reductase 1
Gene Aliases
B5R.1, B5R1, B5R2, NQO3A2, humb5R2
UniGene
Hs.334832
Gene Chromosome Location
Chr.1: 202961873-202967276 on Build GRCh38
Transcripts
NM_016243.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678098 c.379G>C p.E127Q Chr.1: 202965467-202965467 on Build GRCh38
4768001 c.414G>A p.L138L Chr.1: 202965432-202965432 on Build GRCh38
3803504 c.390G>T p.K130N Chr.1: 202965456-202965456 on Build GRCh38
5969077 c.432G>A p.V144V Chr.1: 202965414-202965414 on Build GRCh38
4027125 c.464A>G p.Y155C Chr.1: 202965382-202965382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770048767 Chr.1: 202965402-202965402 on Build GRCh38
rs780286898 Chr.1: 202965479-202965479 on Build GRCh38
rs770705433 Chr.1: 202965516-202965517 on Build GRCh38
rs753718367 Chr.1: 202965363-202965363 on Build GRCh38
rs748682995 Chr.1: 202965496-202965496 on Build GRCh38
rs769938125 Chr.1: 202965476-202965476 on Build GRCh38
rs754811239 Chr.1: 202965450-202965450 on Build GRCh38
rs775782124 Chr.1: 202965406-202965406 on Build GRCh38
rs138315237 Chr.1: 202965381-202965381 on Build GRCh38
rs766155049 Chr.1: 202965444-202965444 on Build GRCh38
rs1053230726 Chr.1: 202965554-202965554 on Build GRCh38
rs779965923 Chr.1: 202965347-202965347 on Build GRCh38
rs774617892 Chr.1: 202965494-202965494 on Build GRCh38
rs574520357 Chr.1: 202965505-202965505 on Build GRCh38
rs201820857 Chr.1: 202965463-202965463 on Build GRCh38
rs200519288 Chr.1: 202965483-202965483 on Build GRCh38
rs111343519 Chr.1: 202965369-202965369 on Build GRCh38
rs772787951 Chr.1: 202965501-202965501 on Build GRCh38
rs562894187 Chr.1: 202965413-202965413 on Build GRCh38
rs768976568 Chr.1: 202965407-202965407 on Build GRCh38
rs753540107 Chr.1: 202965448-202965448 on Build GRCh38
rs749230834 Chr.1: 202965387-202965387 on Build GRCh38
rs759156257 Chr.1: 202965536-202965536 on Build GRCh38
rs771025199 Chr.1: 202965388-202965388 on Build GRCh38
rs376157428 Chr.1: 202965416-202965416 on Build GRCh38
rs778626674 Chr.1: 202965461-202965461 on Build GRCh38
rs531573364 Chr.1: 202965423-202965423 on Build GRCh38
rs767221107 Chr.1: 202965331-202965331 on Build GRCh38
rs760383095 Chr.1: 202965432-202965432 on Build GRCh38
rs781223669 Chr.1: 202965475-202965475 on Build GRCh38
rs200115677 Chr.1: 202965529-202965529 on Build GRCh38
rs1022456347 Chr.1: 202965380-202965380 on Build GRCh38
rs767132971 Chr.1: 202965419-202965419 on Build GRCh38
rs781312449 Chr.1: 202965395-202965395 on Build GRCh38
rs367774238 Chr.1: 202965396-202965396 on Build GRCh38
rs772761478 Chr.1: 202965499-202965499 on Build GRCh38
rs112698958 Chr.1: 202965368-202965368 on Build GRCh38
rs989849089 Chr.1: 202965372-202965372 on Build GRCh38
rs760264121 Chr.1: 202965514-202965514 on Build GRCh38
rs1045566686 Chr.1: 202965510-202965510 on Build GRCh38
rs746997961 Chr.1: 202965383-202965383 on Build GRCh38
rs972504280 Chr.1: 202965467-202965467 on Build GRCh38
rs758368982 Chr.1: 202965471-202965471 on Build GRCh38
rs560661840 Chr.1: 202965346-202965346 on Build GRCh38
rs750086969 Chr.1: 202965339-202965339 on Build GRCh38
rs766085723 Chr.1: 202965515-202965515 on Build GRCh38
rs10920536 Chr.1: 202965480-202965480 on Build GRCh38
rs777913054 Chr.1: 202965342-202965342 on Build GRCh38

View Full Product Details