Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89863416-89863653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8AP2
Entrez Gene ID
9994
Gene Name
caspase 8 associated protein 2
Gene Aliases
CED-4, FLASH, RIP25
UniGene
Hs.558218
Gene Chromosome Location
Chr.6: 89829900-89874436 on Build GRCh38
Transcripts
NM_001137667.1 NM_001137668.1 NM_012115.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286203 c.1750G>A p.D584N Chr.6: 89863459-89863459 on Build GRCh38
484450 c.1916T>C p.L639P Chr.6: 89863625-89863625 on Build GRCh38
6243029 c.1721T>C p.V574A Chr.6: 89863430-89863430 on Build GRCh38
3630512 c.1819C>T p.P607S Chr.6: 89863528-89863528 on Build GRCh38
4597750 c.1751A>C p.D584A Chr.6: 89863460-89863460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774468832 Chr.6: 89863602-89863602 on Build GRCh38
rs554059519 Chr.6: 89863597-89863597 on Build GRCh38
rs267601161 Chr.6: 89863465-89863465 on Build GRCh38
rs748373215 Chr.6: 89863576-89863576 on Build GRCh38
rs370562618 Chr.6: 89863499-89863499 on Build GRCh38
rs781594185 Chr.6: 89863574-89863574 on Build GRCh38
rs777846436 Chr.6: 89863585-89863585 on Build GRCh38
rs749886206 Chr.6: 89863550-89863550 on Build GRCh38
rs368459977 Chr.6: 89863579-89863579 on Build GRCh38
rs763920400 Chr.6: 89863511-89863511 on Build GRCh38
rs771941874 Chr.6: 89863477-89863477 on Build GRCh38
rs746943629 Chr.6: 89863599-89863599 on Build GRCh38
rs761514531 Chr.6: 89863521-89863521 on Build GRCh38
rs772029793 Chr.6: 89863595-89863595 on Build GRCh38
rs377095494 Chr.6: 89863449-89863449 on Build GRCh38
rs764906422 Chr.6: 89863546-89863546 on Build GRCh38
rs748218107 Chr.6: 89863615-89863615 on Build GRCh38
rs772869571 Chr.6: 89863624-89863624 on Build GRCh38
rs375044202 Chr.6: 89863577-89863577 on Build GRCh38
rs776481223 Chr.6: 89863603-89863603 on Build GRCh38
rs749320094 Chr.6: 89863588-89863588 on Build GRCh38
rs372157483 Chr.6: 89863625-89863625 on Build GRCh38
rs755334863 Chr.6: 89863572-89863572 on Build GRCh38
rs534274290 Chr.6: 89863466-89863466 on Build GRCh38
rs760348860 Chr.6: 89863501-89863501 on Build GRCh38
rs751983423 Chr.6: 89863562-89863562 on Build GRCh38
rs570121757 Chr.6: 89863446-89863446 on Build GRCh38
rs773280330 Chr.6: 89863464-89863464 on Build GRCh38
rs753483066 Chr.6: 89863517-89863517 on Build GRCh38
rs780657241 Chr.6: 89863558-89863558 on Build GRCh38
rs758987098 Chr.6: 89863557-89863557 on Build GRCh38

View Full Product Details