Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63323687-63323926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD3
Entrez Gene ID
27022
Gene Name
forkhead box D3
Gene Aliases
AIS1, Genesis, HFH2, VAMAS2
UniGene
Hs.546573
Gene Chromosome Location
Chr.1: 63323059-63325126 on Build GRCh38
Transcripts
NM_012183.2 NR_121637.1

Gene Symbol
FOXD3-AS1
Entrez Gene ID
100996301
Gene Name
FOXD3 antisense RNA 1 (head to head)
Gene Aliases
pasFOXD3
UniGene
Gene Chromosome Location
Chr.1: 63320884-63324441 on Build GRCh38
Transcripts
NM_012183.2 NR_121637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368632 c.715C>A p.R239S Chr.1: 63323773-63323773 on Build GRCh38
6005811 c.754A>T p.M252L Chr.1: 63323812-63323812 on Build GRCh38
6435746 c.787G>A p.A263T Chr.1: 63323845-63323845 on Build GRCh38
6434719 c.721C>A p.Q241K Chr.1: 63323779-63323779 on Build GRCh38
6428498 c.757A>G p.M253V Chr.1: 63323815-63323815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141605755 Chr.1: 63323712-63323712 on Build GRCh38
rs761492867 Chr.1: 63323820-63323820 on Build GRCh38
rs770035947 Chr.1: 63323812-63323812 on Build GRCh38
rs1035389772 Chr.1: 63323830-63323830 on Build GRCh38
rs781209290 Chr.1: 63323878-63323878 on Build GRCh38
rs144838490 Chr.1: 63323752-63323752 on Build GRCh38
rs200810432 Chr.1: 63323807-63323807 on Build GRCh38
rs764763503 Chr.1: 63323860-63323860 on Build GRCh38
rs541120234 Chr.1: 63323908-63323908 on Build GRCh38
rs767405542 Chr.1: 63323829-63323829 on Build GRCh38
rs765929377 Chr.1: 63323854-63323854 on Build GRCh38
rs374869492 Chr.1: 63323802-63323802 on Build GRCh38
rs779367590 Chr.1: 63323784-63323784 on Build GRCh38
rs1024977459 Chr.1: 63323840-63323840 on Build GRCh38
rs747622091 Chr.1: 63323799-63323799 on Build GRCh38
rs769912149 Chr.1: 63323884-63323884 on Build GRCh38
rs369566771 Chr.1: 63323813-63323813 on Build GRCh38
rs923547339 Chr.1: 63323892-63323892 on Build GRCh38
rs758688806 Chr.1: 63323862-63323862 on Build GRCh38
rs779775701 Chr.1: 63323751-63323751 on Build GRCh38
rs760588882 Chr.1: 63323888-63323888 on Build GRCh38
rs754633262 Chr.1: 63323857-63323857 on Build GRCh38
rs745544695 Chr.1: 63323812-63323814 on Build GRCh38
rs1047985255 Chr.1: 63323895-63323895 on Build GRCh38
rs910628448 Chr.1: 63323876-63323876 on Build GRCh38
rs745916627 Chr.1: 63323879-63323879 on Build GRCh38
rs778117598 Chr.1: 63323796-63323796 on Build GRCh38
rs770933954 Chr.1: 63323804-63323804 on Build GRCh38
rs771695051 Chr.1: 63323898-63323898 on Build GRCh38
rs755670470 Chr.1: 63323733-63323733 on Build GRCh38
rs373332012 Chr.1: 63323724-63323724 on Build GRCh38
rs979931976 Chr.1: 63323875-63323875 on Build GRCh38
rs773073356 Chr.1: 63323900-63323900 on Build GRCh38
rs147290223 Chr.1: 63323808-63323808 on Build GRCh38
rs774979471 Chr.1: 63323871-63323871 on Build GRCh38
rs757786681 Chr.1: 63323874-63323874 on Build GRCh38
rs747461169 Chr.1: 63323864-63323864 on Build GRCh38
rs772524720 Chr.1: 63323792-63323792 on Build GRCh38
rs753420481 Chr.1: 63323855-63323855 on Build GRCh38
rs754690288 Chr.1: 63323773-63323773 on Build GRCh38
rs752440666 Chr.1: 63323861-63323861 on Build GRCh38
rs750322647 Chr.1: 63323831-63323831 on Build GRCh38
rs532147202 Chr.1: 63323865-63323866 on Build GRCh38
rs749589976 Chr.1: 63323897-63323897 on Build GRCh38
rs775562720 Chr.1: 63323891-63323891 on Build GRCh38
rs748488056 Chr.1: 63323791-63323791 on Build GRCh38

View Full Product Details