Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143586118-143586391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1D
Entrez Gene ID
1936
Gene Name
eukaryotic translation elongation factor 1 delta
Gene Aliases
EF-1D, EF1D, FP1047
UniGene
Hs.333388
Gene Chromosome Location
Chr.8: 143579697-143597675 on Build GRCh38
Transcripts
NM_001130053.2 NM_001130055.2 NM_001130056.2 NM_001130057.2 NM_001195203.1 NM_001289950.1 NM_001317743.1 NM_001960.4 NM_032378.4 XM_005250820.3 XM_005250823.1 XM_006716519.3 XM_006716520.3 XM_006716522.1 XM_006716523.1 XM_006716524.1 XM_006716525.1 XM_011516905.2 XM_011516906.2 XM_017013168.1 XM_017013169.1 XM_017013170.1 XM_017013171.1 XM_017013172.1 XM_017013173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187884 c.1237C>T p.L413F Chr.8: 143586269-143586269 on Build GRCh38
7187885 c.1237C>T p.L413F Chr.8: 143586269-143586269 on Build GRCh38
5420915 c.1231G>A p.V411M Chr.8: 143586275-143586275 on Build GRCh38
5420914 c.1231G>A p.V411M Chr.8: 143586275-143586275 on Build GRCh38
6651745 c.1278C>A p.S426S Chr.8: 143586228-143586228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774700710 Chr.8: 143586290-143586290 on Build GRCh38
rs373809846 Chr.8: 143586180-143586180 on Build GRCh38
rs995178240 Chr.8: 143586228-143586228 on Build GRCh38
rs781224000 Chr.8: 143586266-143586266 on Build GRCh38
rs751417628 Chr.8: 143586331-143586331 on Build GRCh38
rs1028054990 Chr.8: 143586293-143586293 on Build GRCh38
rs773671363 Chr.8: 143586315-143586315 on Build GRCh38
rs868197356 Chr.8: 143586155-143586155 on Build GRCh38
rs549161091 Chr.8: 143586275-143586275 on Build GRCh38
rs776973558 Chr.8: 143586319-143586319 on Build GRCh38
rs760849438 Chr.8: 143586316-143586316 on Build GRCh38
rs990274631 Chr.8: 143586167-143586167 on Build GRCh38
rs1024830984 Chr.8: 143586281-143586281 on Build GRCh38
rs749699322 Chr.8: 143586194-143586194 on Build GRCh38
rs367863794 Chr.8: 143586200-143586200 on Build GRCh38
rs751616584 Chr.8: 143586256-143586256 on Build GRCh38
rs371789832 Chr.8: 143586280-143586280 on Build GRCh38
rs772590863 Chr.8: 143586312-143586312 on Build GRCh38
rs766031138 Chr.8: 143586318-143586318 on Build GRCh38
rs377645336 Chr.8: 143586183-143586183 on Build GRCh38
rs769131230 Chr.8: 143586289-143586289 on Build GRCh38
rs932821643 Chr.8: 143586171-143586171 on Build GRCh38
rs759457282 Chr.8: 143586342-143586345 on Build GRCh38
rs562856798 Chr.8: 143586206-143586206 on Build GRCh38
rs990218377 Chr.8: 143586335-143586335 on Build GRCh38
rs767944471 Chr.8: 143586208-143586208 on Build GRCh38
rs748452661 Chr.8: 143586301-143586301 on Build GRCh38
rs143904844 Chr.8: 143586285-143586285 on Build GRCh38
rs199981785 Chr.8: 143586178-143586178 on Build GRCh38
rs752561426 Chr.8: 143586173-143586173 on Build GRCh38
rs1020791747 Chr.8: 143586239-143586239 on Build GRCh38
rs766655388 Chr.8: 143586216-143586216 on Build GRCh38
rs769507102 Chr.8: 143586192-143586192 on Build GRCh38
rs965625390 Chr.8: 143586255-143586255 on Build GRCh38
rs761658589 Chr.8: 143586338-143586338 on Build GRCh38
rs551982323 Chr.8: 143586191-143586191 on Build GRCh38
rs201204853 Chr.8: 143586325-143586325 on Build GRCh38
rs760624568 Chr.8: 143586211-143586211 on Build GRCh38
rs567061751 Chr.8: 143586276-143586276 on Build GRCh38
rs776474992 Chr.8: 143586317-143586318 on Build GRCh38
rs953784888 Chr.8: 143586303-143586303 on Build GRCh38
rs758088185 Chr.8: 143586175-143586175 on Build GRCh38
rs779802689 Chr.8: 143586282-143586282 on Build GRCh38
rs746924680 Chr.8: 143586179-143586179 on Build GRCh38
rs775412341 Chr.8: 143586240-143586240 on Build GRCh38
rs371573684 Chr.8: 143586264-143586264 on Build GRCh38
rs139903179 Chr.8: 143586225-143586225 on Build GRCh38
rs759678027 Chr.8: 143586322-143586322 on Build GRCh38
rs966602662 Chr.8: 143586292-143586292 on Build GRCh38
rs201983418 Chr.8: 143586235-143586235 on Build GRCh38
rs770893820 Chr.8: 143586318-143586318 on Build GRCh38
rs527909068 Chr.8: 143586271-143586271 on Build GRCh38
rs912826205 Chr.8: 143586195-143586195 on Build GRCh38
rs202158398 Chr.8: 143586199-143586199 on Build GRCh38
rs369078847 Chr.8: 143586168-143586168 on Build GRCh38
rs150824078 Chr.8: 143586219-143586219 on Build GRCh38
rs533479346 Chr.8: 143586209-143586209 on Build GRCh38

View Full Product Details