Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84554814-84555057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBS
Entrez Gene ID
1486
Gene Name
chitobiase
Gene Aliases
CTB
UniGene
Hs.513557
Gene Chromosome Location
Chr.1: 84553121-84574480 on Build GRCh38
Transcripts
NM_004388.2 NM_001310156.1 XM_011542514.2

Gene Symbol
SPATA1
Entrez Gene ID
100505741
Gene Name
spermatogenesis associated 1
Gene Aliases
SP-2, SPAP1
UniGene
Hs.584956
Gene Chromosome Location
Chr.1: 84506301-84556495 on Build GRCh38
Transcripts
NM_004388.2 NM_001310156.1 XM_011542514.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2243687 c.1125G>A p.W375* Chr.1: 84555032-84555032 on Build GRCh38
5899814 c.1114G>A p.E372K Chr.1: 84555043-84555043 on Build GRCh38
1249364 c.1157G>A p.*386* Chr.1: 84555000-84555000 on Build GRCh38
1249365 c.1152G>A p.Q384Q Chr.1: 84555005-84555005 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs11801891 Chr.1: 84554954-84554954 on Build GRCh38
rs774511903 Chr.1: 84555009-84555009 on Build GRCh38
rs754652720 Chr.1: 84554991-84554991 on Build GRCh38
rs566173961 Chr.1: 84554861-84554861 on Build GRCh38
rs1047754724 Chr.1: 84554917-84554917 on Build GRCh38
rs111674297 Chr.1: 84555015-84555015 on Build GRCh38
rs138201181 Chr.1: 84555000-84555000 on Build GRCh38
rs937152325 Chr.1: 84554873-84554873 on Build GRCh38
rs183916139 Chr.1: 84554947-84554947 on Build GRCh38
rs760513098 Chr.1: 84554981-84554985 on Build GRCh38
rs985142951 Chr.1: 84554840-84554840 on Build GRCh38
rs910725200 Chr.1: 84554881-84554881 on Build GRCh38
rs990096420 Chr.1: 84554892-84554892 on Build GRCh38
rs763056916 Chr.1: 84554992-84554992 on Build GRCh38
rs748114267 Chr.1: 84554936-84554936 on Build GRCh38
rs775834132 Chr.1: 84554971-84554971 on Build GRCh38
rs1037932172 Chr.1: 84554886-84554886 on Build GRCh38
rs889980771 Chr.1: 84554981-84554981 on Build GRCh38
rs775151391 Chr.1: 84554950-84554950 on Build GRCh38
rs193181499 Chr.1: 84554901-84554901 on Build GRCh38
rs943518172 Chr.1: 84554885-84554885 on Build GRCh38
rs746203845 Chr.1: 84554958-84554958 on Build GRCh38
rs945962764 Chr.1: 84554929-84554929 on Build GRCh38
rs753793114 Chr.1: 84555031-84555031 on Build GRCh38
rs1043493 Chr.1: 84554966-84554966 on Build GRCh38
rs534701919 Chr.1: 84554900-84554900 on Build GRCh38
rs763997358 Chr.1: 84555020-84555021 on Build GRCh38
rs34160527 Chr.1: 84555012-84555012 on Build GRCh38

View Full Product Details