Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36210506-36210773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTA
Entrez Gene ID
1211
Gene Name
clathrin light chain A
Gene Aliases
LCA
UniGene
Hs.522114
Gene Chromosome Location
Chr.9: 36190856-36212062 on Build GRCh38
Transcripts
NM_001076677.2 NM_001184760.1 NM_001184761.1 NM_001184762.1 NM_001311203.1 NM_001311204.1 NM_001311205.1 NM_001311206.1 NM_001833.3 NM_007096.3 NR_132349.1 XM_017014257.1 XM_017014258.1

SNP Detail

SNP ID Chromosome Location
rs781739523 Chr.9: 36210675-36210675 on Build GRCh38
rs751965637 Chr.9: 36210639-36210639 on Build GRCh38
rs770506915 Chr.9: 36210606-36210610 on Build GRCh38
rs751999559 Chr.9: 36210536-36210536 on Build GRCh38
rs768131788 Chr.9: 36210601-36210601 on Build GRCh38
rs201366938 Chr.9: 36210598-36210598 on Build GRCh38
rs375414823 Chr.9: 36210660-36210660 on Build GRCh38
rs970551483 Chr.9: 36210681-36210681 on Build GRCh38
rs953666293 Chr.9: 36210716-36210716 on Build GRCh38
rs912063124 Chr.9: 36210730-36210730 on Build GRCh38
rs546548415 Chr.9: 36210587-36210587 on Build GRCh38
rs760665781 Chr.9: 36210583-36210583 on Build GRCh38
rs369930011 Chr.9: 36210709-36210709 on Build GRCh38
rs775246203 Chr.9: 36210575-36210575 on Build GRCh38
rs527361966 Chr.9: 36210750-36210750 on Build GRCh38
rs776426291 Chr.9: 36210686-36210689 on Build GRCh38
rs569628041 Chr.9: 36210691-36210692 on Build GRCh38
rs776697528 Chr.9: 36210589-36210589 on Build GRCh38
rs766906261 Chr.9: 36210638-36210638 on Build GRCh38
rs956055049 Chr.9: 36210693-36210693 on Build GRCh38
rs1026884243 Chr.9: 36210530-36210530 on Build GRCh38
rs200405223 Chr.9: 36210617-36210617 on Build GRCh38
rs759976755 Chr.9: 36210544-36210544 on Build GRCh38
rs1014999746 Chr.9: 36210554-36210554 on Build GRCh38
rs761741991 Chr.9: 36210597-36210597 on Build GRCh38
rs763210801 Chr.9: 36210632-36210632 on Build GRCh38
rs548446012 Chr.9: 36210728-36210728 on Build GRCh38
rs753415697 Chr.9: 36210680-36210680 on Build GRCh38
rs753286788 Chr.9: 36210749-36210749 on Build GRCh38
rs1028044584 Chr.9: 36210685-36210685 on Build GRCh38
rs977169487 Chr.9: 36210739-36210739 on Build GRCh38
rs183321849 Chr.9: 36210654-36210654 on Build GRCh38
rs372212225 Chr.9: 36210717-36210717 on Build GRCh38
rs188261514 Chr.9: 36210738-36210738 on Build GRCh38
rs778459238 Chr.9: 36210701-36210701 on Build GRCh38
rs745559191 Chr.9: 36210708-36210708 on Build GRCh38
rs756686181 Chr.9: 36210691-36210691 on Build GRCh38

View Full Product Details