Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171891410-171891675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4969211 c.640C>T p.R214* Chr.5: 171891577-171891577 on Build GRCh38
3674392 c.740A>G p.N247S Chr.5: 171891477-171891477 on Build GRCh38
420787 c.668G>A p.R223H Chr.5: 171891549-171891549 on Build GRCh38
50194 c.780_788delTTCTATTAA p.N260_I262 Chr.5: 171891468-171891476 on Build GRCh38
3674391 c.779A>G p.N260S Chr.5: 171891477-171891477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779901224 Chr.5: 171891584-171891584 on Build GRCh38
rs149421146 Chr.5: 171891611-171891611 on Build GRCh38
rs771934811 Chr.5: 171891577-171891577 on Build GRCh38
rs774792552 Chr.5: 171891627-171891627 on Build GRCh38
rs151338544 Chr.5: 171891448-171891448 on Build GRCh38
rs752235888 Chr.5: 171891511-171891511 on Build GRCh38
rs965198670 Chr.5: 171891630-171891630 on Build GRCh38
rs767949795 Chr.5: 171891509-171891509 on Build GRCh38
rs374124470 Chr.5: 171891454-171891454 on Build GRCh38
rs773335882 Chr.5: 171891447-171891447 on Build GRCh38
rs770043768 Chr.5: 171891601-171891601 on Build GRCh38
rs746846429 Chr.5: 171891585-171891585 on Build GRCh38
rs778952501 Chr.5: 171891554-171891554 on Build GRCh38
rs753373360 Chr.5: 171891527-171891527 on Build GRCh38
rs755800175 Chr.5: 171891515-171891515 on Build GRCh38
rs762106852 Chr.5: 171891443-171891443 on Build GRCh38
rs765162878 Chr.5: 171891446-171891446 on Build GRCh38
rs745819881 Chr.5: 171891576-171891576 on Build GRCh38
rs747997841 Chr.5: 171891525-171891526 on Build GRCh38
rs755297318 Chr.5: 171891496-171891496 on Build GRCh38
rs756863609 Chr.5: 171891539-171891539 on Build GRCh38
rs762989900 Chr.5: 171891612-171891612 on Build GRCh38
rs766848779 Chr.5: 171891464-171891464 on Build GRCh38
rs752102246 Chr.5: 171891488-171891488 on Build GRCh38
rs545335363 Chr.5: 171891640-171891640 on Build GRCh38
rs771032181 Chr.5: 171891617-171891617 on Build GRCh38
rs367867781 Chr.5: 171891462-171891462 on Build GRCh38
rs777359019 Chr.5: 171891518-171891518 on Build GRCh38
rs767710499 Chr.5: 171891637-171891637 on Build GRCh38
rs760052776 Chr.5: 171891634-171891634 on Build GRCh38

View Full Product Details