Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 100061333-100061562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN5
Entrez Gene ID
53942
Gene Name
contactin 5
Gene Aliases
HNB-2s, NB-2
UniGene
Hs.656783
Gene Chromosome Location
Chr.11: 99020904-100358885 on Build GRCh38
Transcripts
NM_001243270.1 NM_001243271.1 NM_014361.3 NM_175566.2 XM_011542871.1 XM_011542873.1 XM_017017926.1 XM_017017927.1 XM_017017928.1 XM_017017929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4629859 c.1108A>G p.R370G Chr.11: 100061339-100061339 on Build GRCh38
6070723 c.1142G>T p.R381L Chr.11: 100061373-100061373 on Build GRCh38
3454902 c.1127G>A p.G376E Chr.11: 100061358-100061358 on Build GRCh38
6070721 c.1147C>A p.Q383K Chr.11: 100061378-100061378 on Build GRCh38
1721720 c.1136C>T p.S379F Chr.11: 100061367-100061367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200696545 Chr.11: 100061378-100061378 on Build GRCh38
rs201982881 Chr.11: 100061356-100061356 on Build GRCh38
rs1042656506 Chr.11: 100061477-100061477 on Build GRCh38
rs778948748 Chr.11: 100061383-100061383 on Build GRCh38
rs753642668 Chr.11: 100061460-100061460 on Build GRCh38
rs866845091 Chr.11: 100061367-100061367 on Build GRCh38
rs763452468 Chr.11: 100061425-100061425 on Build GRCh38
rs763618297 Chr.11: 100061437-100061437 on Build GRCh38
rs774916419 Chr.11: 100061431-100061431 on Build GRCh38
rs945653260 Chr.11: 100061476-100061476 on Build GRCh38
rs745817608 Chr.11: 100061387-100061387 on Build GRCh38
rs1034240514 Chr.11: 100061520-100061520 on Build GRCh38
rs200717963 Chr.11: 100061355-100061355 on Build GRCh38
rs1012287781 Chr.11: 100061524-100061524 on Build GRCh38
rs374613997 Chr.11: 100061461-100061461 on Build GRCh38
rs772215026 Chr.11: 100061394-100061394 on Build GRCh38
rs767026275 Chr.11: 100061354-100061354 on Build GRCh38
rs903027207 Chr.11: 100061449-100061449 on Build GRCh38
rs780350642 Chr.11: 100061408-100061408 on Build GRCh38
rs3758923 Chr.11: 100061414-100061414 on Build GRCh38
rs148837184 Chr.11: 100061430-100061430 on Build GRCh38
rs748422149 Chr.11: 100061416-100061416 on Build GRCh38
rs770165260 Chr.11: 100061420-100061420 on Build GRCh38
rs200128316 Chr.11: 100061371-100061371 on Build GRCh38
rs574072302 Chr.11: 100061516-100061516 on Build GRCh38
rs768808972 Chr.11: 100061410-100061410 on Build GRCh38
rs777608054 Chr.11: 100061373-100061373 on Build GRCh38
rs1023643734 Chr.11: 100061526-100061526 on Build GRCh38
rs893882228 Chr.11: 100061523-100061523 on Build GRCh38
rs200941149 Chr.11: 100061432-100061432 on Build GRCh38
rs760349613 Chr.11: 100061434-100061434 on Build GRCh38
rs753620493 Chr.11: 100061376-100061376 on Build GRCh38
rs904072643 Chr.11: 100061510-100061510 on Build GRCh38

View Full Product Details