Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31552968-31553241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC129
Entrez Gene ID
223075
Gene Name
coiled-coil domain containing 129
Gene Aliases
-
UniGene
Hs.224269
Gene Chromosome Location
Chr.7: 31514071-31658720 on Build GRCh38
Transcripts
NM_001257967.1 NM_001257968.1 NM_194300.3 NR_047565.1 XM_011515213.1 XM_017011871.1 XM_017011872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5546255 c.101G>A p.G34E Chr.7: 31553047-31553047 on Build GRCh38
3374609 c.89C>T p.A30V Chr.7: 31553113-31553113 on Build GRCh38
1736753 c.44G>A p.G15D Chr.7: 31553068-31553068 on Build GRCh38
6497623 c.102G>T p.L34L Chr.7: 31553126-31553126 on Build GRCh38
746167 c.162G>T p.L54L Chr.7: 31553186-31553186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7811042 Chr.7: 31553119-31553119 on Build GRCh38
rs1045901476 Chr.7: 31553081-31553081 on Build GRCh38
rs771087429 Chr.7: 31553114-31553114 on Build GRCh38
rs765700053 Chr.7: 31553202-31553202 on Build GRCh38
rs763704398 Chr.7: 31553064-31553064 on Build GRCh38
rs373006439 Chr.7: 31553209-31553209 on Build GRCh38
rs368587451 Chr.7: 31553085-31553085 on Build GRCh38
rs749588865 Chr.7: 31553082-31553083 on Build GRCh38
rs780268733 Chr.7: 31553042-31553042 on Build GRCh38
rs769649044 Chr.7: 31553029-31553029 on Build GRCh38
rs868287284 Chr.7: 31553179-31553179 on Build GRCh38
rs112921112 Chr.7: 31553166-31553166 on Build GRCh38
rs369351532 Chr.7: 31553199-31553199 on Build GRCh38
rs751194014 Chr.7: 31553086-31553086 on Build GRCh38
rs143690079 Chr.7: 31553122-31553122 on Build GRCh38
rs779424437 Chr.7: 31553110-31553110 on Build GRCh38
rs759480206 Chr.7: 31553025-31553025 on Build GRCh38
rs746850092 Chr.7: 31552994-31552994 on Build GRCh38
rs114739660 Chr.7: 31553063-31553063 on Build GRCh38
rs376118062 Chr.7: 31553178-31553178 on Build GRCh38
rs762410735 Chr.7: 31553127-31553127 on Build GRCh38
rs534138807 Chr.7: 31553149-31553149 on Build GRCh38
rs550681504 Chr.7: 31553111-31553111 on Build GRCh38
rs147399119 Chr.7: 31553105-31553105 on Build GRCh38
rs756090521 Chr.7: 31553000-31553003 on Build GRCh38
rs774891627 Chr.7: 31553060-31553060 on Build GRCh38
rs371511395 Chr.7: 31553150-31553150 on Build GRCh38
rs750792790 Chr.7: 31553113-31553113 on Build GRCh38
rs148146765 Chr.7: 31552991-31552991 on Build GRCh38
rs374470226 Chr.7: 31553112-31553112 on Build GRCh38
rs372303441 Chr.7: 31553123-31553123 on Build GRCh38
rs1036525081 Chr.7: 31553174-31553174 on Build GRCh38
rs368232809 Chr.7: 31553013-31553013 on Build GRCh38
rs371959143 Chr.7: 31553092-31553092 on Build GRCh38
rs780222183 Chr.7: 31553205-31553205 on Build GRCh38
rs566678979 Chr.7: 31553139-31553139 on Build GRCh38
rs776603611 Chr.7: 31553018-31553018 on Build GRCh38
rs754275371 Chr.7: 31553183-31553183 on Build GRCh38
rs565253184 Chr.7: 31553096-31553096 on Build GRCh38

View Full Product Details